Search
Search

ACVR1| Gene Symbol: ACVR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105.4 | NP_001096.1 | 5 | 814 | ||
| NM_001111067.2 | NP_001104537.1 | 5 | 633 | |||
| XM_005246939.3 | XP_005246996.1 | 5 | 685 | |||
| XM_005246940.3 | XP_005246997.1 | 5 | 653 | |||
| XM_006712825.3 | XP_006712888.1 | 5 | 766 | |||
| XM_011512106.2 | XP_011510408.1 | 6 | 733 | |||
| XM_011512107.2 | XP_011510409.1 | 5 | 705 | |||
| XM_011512108.2 | XP_011510410.1 | 5 | 2112 | |||
| GenBank mRNA | AK297765.1 | - | 5 | 601 | ||
| AK313630.1 | - | 5 | 609 | |||
| BC033867.1 | - | 5 | 814 | 5209711 | ||
| BC036748.1 | - | 5 | 814 | |||
| KU177885.1 | - | 3 | 384 | |||
| KU177886.1 | - | 3 | 384 | |||
| KU177887.1 | - | 3 | 384 | |||
| L02911.1 | - | 6 | 724 | |||
| Z22534.1 | - | 4 | 487 | |||