Search
Search

LOC642929| Gene Symbol: LOC642929 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027472.1 | 2 | 361 | |||
| GenBank mRNA | DQ786189.1 | - | 2 | 360 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BX096823.1 | - | 2 | 386 | Chr. 9 - 66162117 - 66164929 [-] on Build GRCh38 | |