Search
Search

SCN1A| Gene Symbol: SCN1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011511602.2 | XP_011509904.1 | 1 | 178 | ||
| XM_011511605.2 | XP_011509907.1 | 1 | 186 | |||
| XM_011511606.2 | XP_011509908.1 | 1 | 178 | |||
| XM_017004644.1 | XP_016860133.1 | 1 | 178 | |||
| XM_017004645.1 | XP_016860134.1 | 1 | 186 | |||
| XM_017004646.1 | XP_016860135.1 | 1 | 192 | |||
| XM_017004647.1 | XP_016860136.1 | 1 | 185 | |||
| XM_017004650.1 | XP_016860139.1 | 1 | 180 | |||
| XM_017004652.1 | XP_016860141.1 | 1 | 173 | |||
| XM_017004653.1 | XP_016860142.1 | 1 | 191 | |||
| XM_017004654.1 | XP_016860143.1 | 1 | 202 | |||
| GenBank mRNA | DQ993523.1 | - | 1 | 197 | ||