Search
Search

LOC100128006| Gene Symbol: LOC100128006 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_104607.1 | 2 | 2851 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000584772.1 | - | 2 | 2809 | Chr. 17 - 12760140 - 12790242 [-] on Build GRCh38 | |