Search
Search

Taf9b| Gene Symbol: Taf9b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133615.1 | NP_598299.1 | 7 | 607 | ||
| XM_006256994.2 | XP_006257056.1 | 8 | 1073 | |||
| GenBank mRNA | BC090332.1 | - | 7 | 662 | 7193160 | |
| U40188.1 | - | 7 | 607 | |||