Search
Search

Prr5| Gene Symbol: Prr5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012121.1 | NP_001012121.1 | 4, 5 | 500 | ||
| XM_008765722.1 | XP_008763944.1 | 5, 6 | 1596 | |||
| XM_008765723.1 | XP_008763945.1 | 4, 5 | 356 | |||
| XM_008765724.1 | XP_008763946.1 | 4, 5 | 1181 | |||
| XM_008765725.1 | XP_008763947.1 | 5, 6 | 333 | |||
| XM_008765726.1 | XP_008763948.1 | 5, 6 | 328 | |||
| GenBank mRNA | BC090007.1 | - | 4, 5 | 500 | 7323849 | |