Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gabarapl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001044294.1 | NP_001037759.1 | 4 | 482 | ||
GenBank mRNA | BC105627.1 | - | 4 | 482 | 7323087 | |
FQ213425.1 | - | 4 | 394 |