Search
Search

Calml4| Gene Symbol: Calml4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127575.1 | NP_001121047.1 | 3, 4 | 269 | ||
| XM_006243243.2 | XP_006243305.1 | 2, 3 | 219 | |||
| GenBank mRNA | BC158759.1 | - | 3, 4 | 221 | 7099908 | |