Search
Search

COQ7| Gene Symbol: COQ7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001190983.1 | NP_001177912.1 | 2 | 124 | ||
| NM_016138.4 | NP_057222.2 | 2 | 175 | |||
| GenBank mRNA | AF098948.1 | - | 2 | 133 | ||
| AK024291.1 | - | 2 | 156 | |||
| AK225115.1 | - | 2 | 124 | |||
| AK315470.1 | - | 2 | 175 | |||
| AL136647.1 | - | 2 | 127 | |||
| BC003185.1 | - | 2 | 166 | 3503763 | ||
| BC043272.1 | - | 2 | 925 | 5297074 | ||