Search
Search

CAPZA1| Gene Symbol: CAPZA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006135.2 | NP_006126.1 | 8, 9 | 1044 | ||
| XM_017002424.1 | XP_016857913.1 | 8, 9 | 788 | |||
| GenBank mRNA | AK223226.1 | - | 8, 9 | 665 | ||
| AK289654.1 | - | 8, 9 | 687 | |||
| BC000144.1 | - | 8, 9 | 669 | 3047937 | ||
| BT019364.1 | - | 8, 9 | 652 | |||
| BX648738.1 | - | 8, 9 | 1335 | |||
| CR407657.1 | - | 8, 9 | 652 | |||
| CR541819.1 | - | 8, 9 | 652 | |||
| U56637.1 | - | 8, 9 | 652 | |||