Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CAST SIRNA Assays ›
          Gene Symbol:
          CAST
          Gene Name:
          calpastatin
          Gene Aliases:
          BS-17, PLACK
          Chromosome Location:
          Chr.5: 96662037 - 96774683 on Build GRCh38
          UniGene ID:
          Hs.436186
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s2398

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: CAST
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001042440.3 NP_001035905.1 16, 17 1483
          NM_001190442.1 NP_001177371.1 17, 18 1355
          NM_001284212.2 NP_001271141.1 14, 15 1134
          NM_001284213.2 NP_001271142.1 13, 14 1041
          NM_173060.3 NP_775083.1 15, 16 1191
          NR_104285.1 3, 4 347
          XM_006714696.3 XP_006714759.1 19, 20 1497
          XM_006714697.3 XP_006714760.1 18, 19 1514
          XM_006714698.3 XP_006714761.1 17, 18 1475
          XM_006714699.3 XP_006714762.1 18, 19 1438
          XM_006714700.3 XP_006714763.1 17, 18 1457
          XM_006714701.3 XP_006714764.1 17, 18 1473
          XM_006714702.3 XP_006714765.1 17, 18 1349
          XM_006714703.3 XP_006714766.1 16, 17 1434
          XM_006714704.3 XP_006714767.1 17, 18 1382
          XM_006714705.3 XP_006714768.1 16, 17 1391
          XM_006714706.3 XP_006714769.1 16, 17 1309
          XM_006714707.3 XP_006714770.1 16, 17 1260
          XM_006714708.3 XP_006714771.1 15, 16 1220
          XM_006714709.3 XP_006714772.1 15, 16 1183
          XM_006714710.3 XP_006714773.1 14, 15 1138
          XM_006714711.3 XP_006714774.1 14, 15 1183
          XM_006714712.3 XP_006714775.1 13, 14 1130
          XM_006714713.3 XP_006714776.1 3, 4 347
          XM_006714714.3 XP_006714777.1 3, 4 347
          XM_006714715.3 XP_006714778.1 4, 5 1073
          XM_011543654.2 XP_011541956.1 17, 18 1433
          XM_011543655.2 XP_011541957.1 17, 18 1430
          XM_011543656.2 XP_011541958.1 16, 17 1377
          XM_011543657.2 XP_011541959.1 16, 17 1259
          XM_011543658.2 XP_011541960.1 14, 15 1112
          XM_017009911.1 XP_016865400.1 17, 18 1475
          XM_017009912.1 XP_016865401.1 18, 19 1486
          XM_017009913.1 XP_016865402.1 18, 19 1455
          XM_017009914.1 XP_016865403.1 17, 18 1457
          XM_017009915.1 XP_016865404.1 18, 19 1459
          XM_017009916.1 XP_016865405.1 17, 18 1473
          XM_017009917.1 XP_016865406.1 17, 18 1391
          XM_017009918.1 XP_016865407.1 16, 17 1418
          XM_017009919.1 XP_016865408.1 16, 17 1391
          XM_017009920.1 XP_016865409.1 16, 17 1301
          XM_017009921.1 XP_016865410.1 16, 17 1318
          XM_017009922.1 XP_016865411.1 15, 16 1352
          XM_017009923.1 XP_016865412.1 15, 16 1352
          XM_017009924.1 XP_016865413.1 15, 16 1334
          XM_017009925.1 XP_016865414.1 15, 16 1270
          XM_017009926.1 XP_016865415.1 15, 16 1270
          XM_017009927.1 XP_016865416.1 16, 17 1259
          XM_017009928.1 XP_016865417.1 15, 16 1258
          XM_017009929.1 XP_016865418.1 15, 16 1177
          XM_017009930.1 XP_016865419.1 14, 15 1144
          XM_017009931.1 XP_016865420.1 14, 15 1138
          XM_017009932.1 XP_016865421.1 13, 14 1201
          GenBank mRNA AB208816.1 - 17, 18 1319
          AF327443.1 - 12, 13 1033
          AK292289.1 - 3, 4 347
          AK292356.1 - 3, 4 320
          AK296919.1 - 17, 18 1313
          AK297879.1 - 15, 16 1133
          AK298541.1 - 16, 17 1260
          AK299384.1 - 16, 17 1164
          AK299402.1 - 16, 17 1240
          AK300624.1 - 13, 14 1024
          AK302206.1 - 4, 5 347
          AK303836.1 - 9, 10 714
          AK311585.1 - 4, 5 525
          AK311598.1 - 4, 5 401
          AK314290.1 - 14, 15 1136
          AK315146.1 - 14, 15 1135
          AK315673.1 - 3, 4 295
          AL832174.1 - 15, 16 1176
          AL832349.1 - 15, 16 1179
          AY189741.1 - 5, 6 346
          BC013579.1 - 16, 17 1186 3878564
          BC078149.1 - 11, 12 769
          BT009783.1 - 13, 14 968
          BX648397.1 - 14, 15 1085
          D16217.1 - 16, 17 1253
          DB033043.1 - 3, 4 295
          U26724.2 - 7, 8 616
          U31345.1 - 5, 6 363
          U58996.2 - 3, 4 326

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          protein-binding activity modulator protease inhibitor

          Gene Ontology Categories:

          Function(s) Process(es)

          liver development
          egg activation
          brain development
          myoblast fusion
          aging
          negative regulation of endopeptidase activity
          organ regeneration
          myoblast differentiation
          negative regulation of cell cycle arrest
          cell-cell adhesion
          negative regulation of type B pancreatic cell apoptotic process
          protease binding
          endopeptidase inhibitor activity
          protein binding
          calcium-dependent cysteine-type endopeptidase inhibitor activity
          poly(A) RNA binding
          cadherin binding involved in cell-cell adhesion

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-jct7h:80/100.66.72.173:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0