Search
Search

PRR5| Gene Symbol: PRR5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017528.2 | NP_001017528.1 | 9 | 1722 | ||
| NM_001017529.2 | NP_001017529.1 | 8 | 1605 | |||
| NM_001017530.1 | NP_001017530.1 | 8 | 1779 | |||
| NM_001198721.1 | NP_001185650.1 | 10 | 2435 | |||
| NM_015366.3 | NP_056181.2 | 9 | 1896 | |||
| NM_181333.3 | NP_851850.1 | 8 | 1757 | |||
| GenBank mRNA | AF177331.1 | - | 8 | 1701 | ||
| AK000192.1 | - | 4 | 1815 | |||
| AK055848.1 | - | 10 | 2157 | |||
| AK056787.1 | - | 2 | 1843 | |||
| AK091884.1 | - | 9 | 1647 | |||
| BC016921.1 | - | 4 | 2004 | 4413787 | ||