Search
Search

SLAMF8| Gene Symbol: SLAMF8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020125.2 | NP_064510.1 | 3 | 725 | ||
| XM_005245345.2 | XP_005245402.1 | 3 | 764 | |||
| XM_011509775.1 | XP_011508077.1 | 3 | 764 | |||
| XM_011509776.2 | XP_011508078.1 | 3 | 626 | |||
| XM_011509777.2 | XP_011508079.1 | 3 | 764 | |||
| XM_011509778.2 | XP_011508080.1 | 3 | 515 | |||
| XM_011509779.2 | XP_011508081.1 | 2 | 320 | |||
| XM_011509780.2 | XP_011508082.2 | 2 | 332 | |||
| XM_017001844.1 | XP_016857333.1 | 3 | 764 | |||
| XM_017001845.1 | XP_016857334.1 | 3 | 764 | |||
| XM_017001846.1 | XP_016857335.1 | 3 | 612 | |||
| GenBank mRNA | AF146761.1 | - | 3 | 603 | ||
| AK074669.1 | - | 3 | 664 | |||
| BC109194.2 | - | 3 | 541 | 40004601 | ||