Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNX6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021249.3 | NP_067072.3 | 4 | 494 | ||
NM_152233.2 | NP_689419.2 | 5 | 389 | |||
XM_006720224.1 | XP_006720287.1 | 4 | 276 | |||
GenBank mRNA | AF121856.1 | - | 5 | 380 | ||
AK292250.1 | - | 5 | 372 | |||
AK296213.1 | - | 5 | 383 | |||
AK308464.1 | - | 5 | 383 | |||
BC001798.1 | - | 4 | 494 | 3354467 | ||
BG722853.1 | - | 5 | 415 | |||
U83194.1 | - | 5 | 357 |