Search
Search

KATNAL1| Gene Symbol: KATNAL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014380.2 | NP_001014402.1 | 7 | 958 | ||
| NM_032116.4 | NP_115492.1 | 7 | 1000 | |||
| XM_005266574.2 | XP_005266631.1 | 7 | 1142 | |||
| XM_017020791.1 | XP_016876280.1 | 6 | 844 | |||
| XM_017020792.1 | XP_016876281.1 | 6 | 865 | |||
| XM_017020793.1 | XP_016876282.1 | 4 | 1174 | |||
| GenBank mRNA | AK291439.1 | - | 7 | 933 | ||
| AK315724.1 | - | 7 | 937 | |||
| BC000612.2 | - | 7 | 960 | 3347282 | ||