Search
Search

TMEM55B| Gene Symbol: TMEM55B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100814.2 | NP_001094284.1 | 3, 4 | 746 | ||
| NM_144568.3 | NP_653169.2 | 3, 4 | 725 | |||
| GenBank mRNA | AK314021.1 | - | 3, 4 | 726 | ||
| BC002867.1 | - | 3, 4 | 507 | 3940519 | ||
| BC020947.1 | - | 3, 4 | 590 | 4816227 | ||
| BX161490.1 | - | 3, 4 | 591 | |||
| BX248025.1 | - | 3, 4 | 555 | |||