Search
Search

| Gene Symbol: SIGLEC16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002825.2 | 2 | 447 | |||
| GenBank mRNA | AK301031.1 | - | 2 | 447 | ||
| BC039008.1 | - | 2 | 374 | |||
| EU330985.1 | - | 2 | 356 | |||
| Gene Symbol: SIGLEC11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135163.1 | NP_001128635.1 | 2 | 472 | ||
| NM_052884.2 | NP_443116.2 | 2 | 472 | |||
| XM_005258476.3 | XP_005258533.2 | 2 | 413 | |||
| XM_005258477.3 | XP_005258534.2 | 2 | 413 | |||
| GenBank mRNA | AB072038.1 | - | 2 | 391 | ||
| AF337818.1 | - | 2 | 472 | |||
| AY358135.1 | - | 2 | 350 | |||