Search
Search

GANC| Gene Symbol: GANC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198141.2 | NP_937784.2 | 15 | 1982 | ||
| GenBank mRNA | AF525397.1 | - | 11 | 1235 | ||
| AF545045.1 | - | 15 | 1676 | |||
| AF545046.1 | - | 15 | 1676 | |||
| AK074037.1 | - | 15 | 1874 | |||
| BC059406.1 | - | 15 | 1854 | 30330796 | ||
| BC093833.1 | - | 15 | 1699 | 7939678 | ||
| BC112051.1 | - | 15 | 1699 | 8327519 | ||