Search
Search

ANPEP| Gene Symbol: ANPEP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001150.2 | NP_001141.2 | 2 | 503 | ||
| XM_005254892.4 | XP_005254949.1 | 2 | 511 | |||
| XM_011521473.1 | XP_011519775.1 | 2 | 654 | |||
| GenBank mRNA | AB209918.1 | - | 2 | 499 | ||
| BC058928.1 | - | 1 | 304 | 6020902 | ||
| BQ879097.1 | - | 2 | 514 | |||
| M22324.1 | - | 1 | 331 | |||
| X13276.1 | - | 1 | 235 | |||