Search
Search

Krt32| Gene Symbol: Krt32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159374.2 | NP_001152846.2 | 4 | 766 | ||
| GenBank mRNA | AK009595.1 | - | 2 | 93 | ||
| BC117553.1 | - | 4 | 656 | 40104816 | ||
| BC118064.1 | - | 4 | 656 | 40104817 | ||
| X75649.1 | - | 4 | 656 | |||