Search
Search

Ecsit| Gene Symbol: Ecsit | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001253897.1 | NP_001240826.1 | 4 | 479 | ||
| NM_001253898.1 | NP_001240827.1 | 3 | 301 | |||
| NM_012029.2 | NP_036159.1 | 4 | 487 | |||
| GenBank mRNA | AF182510.1 | - | 3 | 500 | ||
| AK007564.1 | - | 4 | 461 | |||
| AK012835.1 | - | 4 | 459 | |||
| AK219052.1 | - | 1 | 25 | |||
| BC004583.1 | - | 3 | 288 | 3587327 | ||
| BC016424.1 | - | 4 | 427 | 4924678 | ||
| BY123162.1 | - | 3 | 281 | |||
| CB589832.1 | - | 2 | 183 | |||