Search
Search

Fut8| Gene Symbol: Fut8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001252614.1 | NP_001239543.1 | 10 | 1546 | ||
| NM_001252615.1 | NP_001239544.1 | 4 | 620 | |||
| NM_001252616.1 | NP_001239545.1 | 3 | 640 | |||
| NM_016893.5 | NP_058589.2 | 9 | 1457 | |||
| GenBank mRNA | AB025198.1 | - | 6 | 1146 | ||
| AK048520.1 | - | 9 | 1595 | |||
| AK051811.1 | - | 9 | 1414 | |||
| AK078788.1 | - | 1 | 161 | |||
| AK134537.1 | - | 9 | 1550 | |||
| AK211417.1 | - | 2 | 181 | |||
| BC010666.1 | - | 10 | 1525 | 3594582 | ||
| EF591874.1 | - | 4 | 620 | |||
| EF591875.1 | - | 3 | 640 | |||