Search
Search

Dctn5| Gene Symbol: Dctn5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021608.3 | NP_067621.3 | 5 | 482 | ||
| GenBank mRNA | AF190795.1 | - | 5 | 453 | ||
| AK010238.1 | - | 5 | 462 | |||
| AK051957.1 | - | 5 | 421 | |||
| AK076100.1 | - | 5 | 455 | |||
| AK076738.1 | - | 5 | 482 | |||
| AK145958.1 | - | 4 | 1433 | |||
| AK146123.1 | - | 5 | 448 | |||
| AK151193.1 | - | 5 | 454 | |||
| AK151876.1 | - | 5 | 454 | |||
| AK168983.1 | - | 5 | 462 | |||
| BC005671.1 | - | 4 | 1456 | 3153175 | ||
| BC010834.1 | - | 5 | 451 | 4235312 | ||
| BC092273.1 | - | 4 | 1435 | |||