Search
Search

LMX1B| Gene Symbol: LMX1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001174146.1 | NP_001167617.1 | 3 | 476 | ||
| NM_001174147.1 | NP_001167618.1 | 3 | 476 | |||
| NM_002316.3 | NP_002307.2 | 3 | 476 | |||
| GenBank mRNA | AF057135.1 | - | 3 | 400 | ||
| BC069588.1 | - | 3 | 395 | |||
| BC069601.1 | - | 3 | 400 | 7262383 | ||
| BC112120.1 | - | 3 | 450 | 8327588 | ||
| BC113491.1 | - | 3 | 450 | 8322543 | ||
| BC143801.1 | - | 3 | 450 | 9052315 | ||
| KU178166.1 | - | 3 | 400 | |||
| KU178167.1 | - | 3 | 400 | |||
| U77457.1 | - | 1 | 118 | |||