Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdcl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026176.3 | NP_080452.2 | 4 | 702 | ||
GenBank mRNA | AK004704.1 | - | 4 | 1415 | ||
AK028668.1 | - | 4 | 592 | |||
AK028814.1 | - | 4 | 592 | |||
AK133747.1 | - | 4 | 612 | |||
AK166983.1 | - | 4 | 592 | |||
BC006578.1 | - | 4 | 602 | 3586612 |