Search
Search

Samd8| Gene Symbol: Samd8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026283.2 | NP_080559.1 | 2 | 386 | ||
| GenBank mRNA | AK006211.1 | - | 2 | 387 | ||
| AK015593.1 | - | 2 | 365 | |||
| AK075749.1 | - | 2 | 337 | |||
| AK147234.1 | - | 2 | 190 | |||
| AK147528.1 | - | 2 | 190 | |||
| BC100476.1 | - | 2 | 401 | 30916247 | ||
| BC116745.1 | - | 2 | 374 | 40126064 | ||
| BC116747.1 | - | 2 | 374 | 40126066 | ||