Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nup35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190179.1 | NP_001177108.1 | 2 | 155 | ||
NM_027091.4 | NP_081367.1 | 2 | 135 | |||
GenBank mRNA | AF411517.1 | - | 2 | 114 | ||
AK009187.1 | - | 2 | 83 | |||
AK089997.1 | - | 2 | 134 | |||
AK136109.1 | - | 2 | 98 | |||
AK145775.1 | - | 2 | 82 | |||
BC048814.1 | - | 2 | 90 | 2647417 | ||
BY272877.1 | - | 2 | 135 | |||
CA560987.1 | - | 2 | 155 | |||
DV653588.1 | - | 2 | 130 |