Search
Search

Cars2| Gene Symbol: Cars2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024248.1 | NP_077210.1 | 8 | 894 | ||
| GenBank mRNA | AK009937.1 | - | 8 | 856 | ||
| AK038968.1 | - | 8 | 904 | |||
| AK041962.1 | - | 8 | 865 | |||
| AK054486.1 | - | 8 | 888 | |||