Search
Search

Rpusd3| Gene Symbol: Rpusd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033204.1 | NP_001028376.1 | 5 | 487 | ||
| GenBank mRNA | AK076749.1 | - | 5 | 488 | ||
| AK133871.1 | - | 5 | 467 | |||
| BC038081.1 | - | 5 | 428 | |||
| BC116825.1 | - | 5 | 487 | 40126144 | ||
| BC116827.1 | - | 5 | 487 | 40126146 | ||
| BC144733.1 | - | 5 | 487 | 9053281 | ||
| BC144734.1 | - | 5 | 487 | |||