Contact Us
ImmunoCAP Phadiatop Test
Help Rule in or out IgE-mediated Allergy
These products may not be cleared for use in your country. Please contact your sales representative for information about specific product availability.
ImmunoCAP™ Phadiatop™ test measures IgE antibodies to a well-balanced mixture of common inhalant allergens and helps rule in or out atopic allergy, a tendency to produce IgE mediated response to protein(s) in the environment. ImmunoCAP Phadiatop Infant, optimized for children younger than 5 years old, includes food allergens that are important for that age group.1-5
These tests can help clinicians:
  • Differentiate IgE-mediated allergic disorders from other allergy-like symptoms.2-4
  • Identify patients who need specific IgE testing with whole allergens to help identify the symptom-provoking allergens and determine appropriate patient management.2-4
ImmunoCAP Phadiatop testing can be performed for any patient
ImmunoCAP Phadiatop testing can be performed with few limitations. Unlike traditional skin-prick testing (SPT), an ImmunoCAP blood test:
  • Can be performed irrespective of a patient’s age, skin condition, medication, symptom, disease
  • activity, pregnancy, and even in early infancy.9-13
  • Poses no risk for anaphylaxis.9,13
  • Is not subject to interference by exogenous biotin supplementation.14
To help identify the symptom-provoking allergens and the quantitative values, it is recommended to retest the sample with the appropriate ImmunoCAP Specific IgE whole allergens.
Nine out of 10 allergic/non-allergic patients correctly classified using ImmunoCAP Phadiatop test
Explore the full ImmunoCAP portfolio.
View Portfolio
Find an instrument to run ImmunoCAP tests.
View Phadia Laboratory Systems
Get scientific information on whole allergens, allergen mixes, and allergen components.
Visit Allergen Encyclopedia
References
1. Directions for use. ImmunoCAP Phadiatop.
2. Eriksson, N.E., Allergy screening with Phadiatop and CAP Phadiatop in combination with a questionnaire in adults with asthma and rhinitis. Allergy, 1990. 45(4): p. 285-92.
3. Fiocchi, A., et al., Differential diagnosis of IgE-mediated allergy in young children with wheezing or eczema symptoms using a single blood test. Ann Allergy Asthma Immunol, 2004. 93(4): p. 328-33.
4. Nilsson C, et al. Pediatr Allergy Immunol. 2012 Mar;23(2):159–65. 
5. Ballardini, N., et al., ImmunoCAP Phadiatop Infant--a new blood test for detecting IgE sensitisation in children at 2 years of age. Allergy, 2006. 61(3): p. 337-43.
6. Williams, P.B., C. Siegel, and J. Portnoy, Efficacy of a single diagnostic test for sensitization to common inhalant allergens. Ann Allergy Asthma Immunol, 2001. 86(2): p. 196-202.
7. Halvorsen, R., et al., Phadiatop infant in the diagnosis of atopy in children with allergy-like symptoms. Int J Pediatr, 2009. 2009: p. 460737.
8. Paganelli, R., et al., Specific IgE antibodies in the diagnosis of atopic disease. Clinical evaluation of a new in vitro test system, UniCAP, in six European allergy clinics. Allergy, 1998. 53(8): p. 763-8.
9. Siles, R.I. and F.H. Hsieh, Allergy blood testing: A practical guide for clinicians. Cleve Clin J Med, 2011. 78(9): p. 585-92.
10. Bonnelykke, K., C.B. Pipper, and H. Bisgaard, Sensitization does not develop in utero. J Allergy Clin Immunol, 2008. 121(3): p. 646-51.
11. Belhocine, W., et al., Total serum tryptase levels are higher in young infants. Pediatr Allergy Immunol, 2011. 22(6): p. 600-7.
12. Bacharier, L.B., et al., Diagnosis and treatment of asthma in childhood: a PRACTALL consensus report. Allergy, 2008. 63(1): p. 5-34.
13. Walsh, J. and N. O'Flynn, Diagnosis and assessment of food allergy in children and young people in primary care and community settings: NICE clinical guideline. The British journal of general practice : the journal of the Royal College of General Practitioners, 2011. 61(588): p. 473-475.
14. ImmunoDiagnostics Statment, No Biotin Interference in ImmunoCAPTM Assays. Uppsala, Jan 31 2018.