Recent searches Clear History
聯絡我們
ImmunoCAP Phadiatop 檢測
幫助納入或排除 IgE 誘發型過敏的可能性
這些產品可能無法進口至您的國家。請聯繫您的銷售代表取得特定產品供貨情形的相關資訊。
ImmunoCAP™ Phadiatop™ 檢測在測量常見吸入性過敏原均勻混合物的 IgE 抗體,能幫助納入和排除異位性過敏的可能性,異位性過敏為一種對環境中蛋白質產生 IgE 誘發型反應之趨勢。ImmunoCAP Phadiatop Infant 針對小於 5 歲之孩童進行了優化,包括對該年齡族群來說相當重要的食物過敏原。1-5
這些檢測可幫助臨床醫師:
  • 鑑別 IgE 誘發型過敏疾患,與其他類似過敏的症狀區別。2-4
  • 辨識出需要以全過敏原進行特異性 IgE 檢測之病患,幫助鑑定誘發症狀之過敏原並判斷適當的病患管理。2-4
ImmunoCAP Phadiatop 檢測可執行於任何病患
ImmunoCAP Phadiatop 檢測可在極少的限制下執行。不同於傳統的皮膚點刺測試 (SPT),ImmunoCAP 血液檢測:
  • 其執行不受限於病患的年齡、皮膚狀況、用藥、症狀、疾病
  • 活動性、懷孕、甚至是早期嬰幼兒。9-13
  • 不會有過敏性休克之風險。9,13
  • 不會受到外源性生物素營養補充品之干擾。14
幫助辨識出誘發症狀的過敏原和定量數值,建議以適當的 ImmunoCAP Specific IgE 全過敏原重新檢測樣本。
10 位過敏/非過敏病患中有九位可利用 ImmunoCAP Phadiatop 檢測出來
探索完整的 ImmunoCAP 產品套組。
檢視產品套組
查看可執行 ImmunoCAP 檢測的儀器。
檢視 Phadia 實驗室系統
取得有關過敏原、過敏原混合物及過敏原成分的科學資訊。
造訪過敏原百科全書
參考資料
1.Directions for use.ImmunoCAP Phadiatop.
2.Eriksson, N.E., Allergy screening with Phadiatop and CAP Phadiatop in combination with a questionnaire in adults with asthma and rhinitis.Allergy, 1990.45(4): p. 285-92.
3.Fiocchi, A., et al., Differential diagnosis of IgE-mediated allergy in young children with wheezing or eczema symptoms using a single blood test.Ann Allergy Asthma Immunol, 2004.93(4): p. 328-33.
4.Nilsson C, et al.Pediatr Allergy Immunol.2012 Mar;23(2):159–65. 
5.Ballardini, N., et al., ImmunoCAP Phadiatop Infant--a new blood test for detecting IgE sensitisation in children at 2 years of age.Allergy, 2006.61(3): p. 337-43.
6.Williams, P.B., C. Siegel, and J. Portnoy, Efficacy of a single diagnostic test for sensitization to common inhalant allergens.Ann Allergy Asthma Immunol, 2001.86(2): p. 196-202.
7.Halvorsen, R., et al., Phadiatop infant in the diagnosis of atopy in children with allergy-like symptoms.Int J Pediatr, 2009.2009: p. 460737.
8.Paganelli, R., et al., Specific IgE antibodies in the diagnosis of atopic disease.Clinical evaluation of a new in vitro test system, UniCAP, in six European allergy clinics.Allergy, 1998.53(8): p. 763-8.
9.Siles, R.I. and F.H.Hsieh, Allergy blood testing: A practical guide for clinicians.Cleve Clin J Med, 2011.78(9): p. 585-92.
10.Bonnelykke, K., C.B.Pipper, and H. Bisgaard, Sensitization does not develop in utero.J Allergy Clin Immunol, 2008.121(3): p. 646-51.
11.Belhocine, W., et al., Total serum tryptase levels are higher in young infants.Pediatr Allergy Immunol, 2011.22(6): p. 600-7.
12.Bacharier, L.B., et al., Diagnosis and treatment of asthma in childhood: a PRACTALL consensus report.Allergy, 2008.63(1): p. 5-34.
13.Walsh, J. and N. O'Flynn, Diagnosis and assessment of food allergy in children and young people in primary care and community settings: NICE clinical guideline.The British journal of general practice : the journal of the Royal College of General Practitioners, 2011.61(588): p. 473-475.
14.ImmunoDiagnostics Statment, No Biotin Interference in ImmunoCAPTM Assays.Uppsala, Jan 31 2018.