protocol
Related Product Information
  • Taq DNA Polymerase
Introduction
Taq DNA Polymerase is isolated from Thermus aquaticus YT1. The enzyme consists of a single polypeptide with a molecular weight of approximately 94 kDa. Taq DNA polymerase is heat-stable and will synthesize DNA at elevated temperatures from single-stranded templates in the presence of a primer.
Kit Size
Component 100 U
500 U
1500 U 5,000 U
Taq DNA Polymerase 20 μl
100 μl
300 μl 1000 μl
10X PCR Buffer, Minus Mg
1.25 ml
2.5 ml
7.5 ml
20 ml
 50 mM Magnesium Chloride
1 ml
1 ml
3 ml
10 ml
Storage Buffer
20 mM Tris-HCl (pH 8.0), 0.1 mM EDTA, 1 mM DTT, 50% (v/v) glycerol, Stabilizers
10X PCR Buffer
200 mM Tris-HCl (pH 8.4), 500 mM KCl The PCR Buffer is supplied as a 10X concentrate and should be diluted for use.
Unit Definition
One unit incorporates 10 nmol of deoxyribonucleotide into acid-precipitable material in 30 minutes at 74°C. Unit assay conditions: 25 mM TAPS (pH 9.3), 50 mM KCl, 2 mM MgCl 2, 1 mM DTT, 0.5 mg/ml activated salmon sperm DNA, 0.2 mM dATP, dCTP, dGTP, dTTP
TOP
Basic PCR Protocol
The following basic protocol serves as a general guideline and a starting point for any PCR amplification. Optimal reaction conditions (incubation times and temperatures, concentration of Taq DNA Polymerase, primers, MgCl 2, and template DNA) vary and need to be optimized. Critical parameters and troubleshooting information are documented in reference 1. PCR reactions should be assembled in a DNA-free environment. Use of "clean" dedicated automatic pipettors and aerosol resistant barrier tips are recommended. Always keep the control DNA and other templates to be amplified isolated from the other components.
  1. Add the following components to a sterile 0.5-ml microcentrifuge tube sitting on ice:
    Component Volume
    Final conc.
    10X PCR buffer minus Mg 10 μl 1X
    10 mM dNTP mixture 2 μl 0.2 mM each
    50 mM MgCl2 3 μl 1.5 nM
    Primer mix (10 μM each) 5 μl 0.5 μM each
    Template DNA  1–20 μl n/a
    Taq DNA Polymerase (5 U/μl)
    0.2–0.5 μl
    1.0–2.5 units
    Autoclaved distilled water
     to 100 μl n/a
  2. Mix contents of tube and overlay with 50 μl of mineral or silicone oil.
  3. Cap tubes and centrifuge briefly to collect the contents to the bottom.
  4. Incubate tubes in a thermal cycler at 94°C for 3 minutes to completely denature the template.
    Perform 25–35 cycles of PCR amplification as follows:
    Denature 94°C for 45 s
    Anneal 55°C for 30 s
    Extend 72°C for 1 min 30 s
  5. Incubate for an additional 10 min at 72°C and maintain the reaction at 4°C. The samples can be stored at –20°C until use.
  6. Analyze the amplification products by agarose gel electrophoresis and visualize by ethidium bromide staining. Use appropriate molecular weight standards.
TOP
"Hot Start" Protocol
In the "hot-start" method, the addition of Taq DNA Polymerase is withheld until the reaction temperature is at 80°C, to ensure high specificity of the products being synthesized.
  1. Add all components as in the Basic PCR Protocol, except for the Taq DNA Polymerase.
  2. Mix contents of tube and overlay with 50 μl of mineral or silicone oil.
  3. Cap tubes and centrifuge briefly to collect the contents to the bottom.
  4. Incubate tubes in a thermal cycler at 94°C for 3 minutes to completely denature the template.
  5. After denaturation at 94°C, maintain the reaction at 80°C.
  6. Add 0.2–0.5 μl of Taq DNA Polymerase (1.0–2.5 U) to each reaction. Be certain to add the enzyme beneath the layer of oil.
  7. Continue with 25–35 cycles of denaturation, annealing and extension as in the Basic PCR Protocol.
TOP
Reference
Innis, M.A., Gelfand, D.H., Sninsky, J.J., and White, T.J., eds. (1990) PCR Protocols: A Guide to Methods and Applications, Academic Press, San Diego, CA.
TOP
MAN0001335      7-Jun-2010