Platinum™ Multiplex PCR Master Mix
Applied Biosystems™

Platinum™ Multiplex PCR Master Mix

The Platinum Multiplex PCR Master Mix is designed specifically for endpoint multiplex PCR.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberNo. of Reactions
44642702000 Reactions
446426850 Reactions
4464269250 Reactions
Catalog number 4464270
Price (USD)
6,410.65
Online exclusive
6,820.00
Save 409.35
Each
No. of Reactions:
2000 Reactions
Request bulk or custom format
Price (USD)
6,410.65
Online exclusive
6,820.00
Save 409.35
Each
Platinum Multiplex PCR Master Mix is a high-specificity, high-throughput solution designed specifically for endpoint multiplex PCR and supports easy multiplexing with minimal optimization.

The performance of the Platinum Multiplex PCR Master Mix over a wide range of amplicon sizes permits the amplification of templates from 50 bp to 2.5 kb, greatly enhancing workflow flexibility. Coupled with its 20-plex capability and absence of primer dimers, it not only provides a high-throughput solution but also boasts high specificity through fewer non-specific primer binding events, resulting in less reaction and primer waste.

Features

  • Amplifies up to 20 amplicons in a single reaction
  • Amplifies products from 50 bp to 2.5 kb
  • Performs multiplex PCR reactions with minimal optimization
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Fidelity (vs. Taq)1X
Hot StartBuilt-In Hot Start
Multiplex CapabilityUp to 20-plex
No. of Reactions2000 Reactions
PolymerasePlatinum Taq DNA Polymerase
Quantity2000 Reactions
Reaction FormatSuperMix or Master Mix
Shipping ConditionDry Ice
Size (Final Product)2.5 kb or less
Volume10 mL
Concentration2X
For Use With (Application)Hot-start PCR, Multiplex PCR
GC-Rich PCR PerformanceLow
Reaction SpeedStandard
Unit SizeEach
Contents & Storage
• Platinum Multiplex PCR Master Mix, 2X (5 x 10 mL)
• GC Enhancer (1 x 10 mL)

Store at –15°C to –25°C prior to opening.
Master mix may be stored at –15°C to –25°C or at 4°C (for up to 30 days). The GC Enhancer must be kept at –15°C to –25°C.

Frequently asked questions (FAQs)

Does the Platinum Multiplex PCR master mix leave A overhangs?

Yes, the Platinum Multiplex PCR master mix contains Platinum Taq DNA Polymerase that leaves A overhangs on PCR products.

What is the difference between Platinum technology and AccuPrime technology?

With Platinum technology, anti-DNA polymerase antibodies bind to the enzyme until the denaturing step at 94 degrees C, when the antibodies degrade. The polymerase is now active and primer extension can occur. AccuPrime Taq combines Platinum Taq (Taq + Platinum antibodies) with proprietary thermostable AccuPrime accessory proteins. The 10X reaction buffer contains the accessory proteins which enhance specific primer-template hybridization during each cycle of PCR.

Can I use Platinum Multiplex PCR Master Mix for amplifying uracil-containing templates?

Yes, Platinum Multiplex PCR Master Mix contains Platinum Taq Polymerase that can incorporate dUTP and amplify uracil-containing templates.