Search
Search

| Assay Name: | bta-miR-181a |
|---|---|
| miRBase Accession Number: | MI0010484 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | AACAUUCAACGCUGUCGGUGAGUU |
| Species: | Bovine, Columba livia, Ophiophagus hannah, Pig |
| Product Type: | mirVana® miRNA inhibitor |
Gene Family ID: | MIPF0000007, mir-181 |
Mature miRNA Sequence: | AACAUUCAACGCUGUCGGUGAGUU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Bovine | bta-miR-181a | MIMAT0003543 |
| Columba livia | cli-miR-181a-5p | MIMAT0038550 |
| Ophiophagus hannah | oha-miR-181a-5p | MIMAT0036757 |
| Pig | ssc-miR-181a | MIMAT0010191 |