Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › MC11451
          Assay Name: bta-miR-181a
          miRBase Accession Number: MI0010484
          miRBase Version: v22.1
          Chromosome Location: -
          Mature miRNA Sequence: AACAUUCAACGCUGUCGGUGAGUU
          Species: Bovine, Columba livia, Ophiophagus hannah, Pig
          Product Type: mirVana® miRNA mimic
          Assay ID MC11451

          Purification
          Size
          Availability Made To Order
          Catalog # 4464066
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000007, mir-181

          Mature miRNA Sequence:

          AACAUUCAACGCUGUCGGUGAGUU

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Bovine bta-miR-181a MIMAT0003543
          Columba livia cli-miR-181a-5p MIMAT0038550
          Ophiophagus hannah oha-miR-181a-5p MIMAT0036757
          Pig ssc-miR-181a MIMAT0010191

          Stem-loop Details



          Bovine

          Stem-loop ID bta-mir-181a-1
          Stem-loop Accession # MI0010484
          Stem-loop Sequence
          UGAGCUCCGAGGUUGCUUCAGUGAACAUUCAACGCUGUCGGUGAGUUUGGAAUUAAAAAUCAAAACCAUCGACCGUUGAUUGUACCCUAUGGCCAACCACCAUCUCCACC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0003543
          miRBase ID: bta-miR-181a
          Mature miRNA Sequence: AACAUUCAACGCUGUCGGUGAGUU
          Chromosome Location: NA

          Stem-loop ID bta-mir-181a-2
          Stem-loop Accession # MI0004757
          Stem-loop Sequence
          UGCCAGGGCCAGGACCCAGUCUUCAGAGGACUCCAAGGAACAUUCAACGCUGUCGGUGAGUUUGGGAUUUGAAAAAACCACCGACCGUUGACUGUACCUUGGGUUCCUUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0003543
          miRBase ID: bta-miR-181a
          Mature miRNA Sequence: AACAUUCAACGCUGUCGGUGAGUU
          Chromosome Location: NA

          Columba livia

          Stem-loop ID cli-mir-181a-1
          Stem-loop Accession # MI0029989
          Stem-loop Sequence
          UGUAGUGGUUGCUUCAGUGAACAUUCAACGCUGUCGGUGAGUUUGGAAUUAAAGUGAAAACCAUCGACCGUUGAUUGUACCCUCCAGCUAACCAUCCUCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038550
          miRBase ID: cli-miR-181a-5p
          Mature miRNA Sequence: AACAUUCAACGCUGUCGGUGAGUU
          Chromosome Location: NA

          Stem-loop ID cli-mir-181a-2
          Stem-loop Accession # MI0029990
          Stem-loop Sequence
          AUCUUUGGAUAGCUUCAGUGAACAUUCAACGCUGUCGGUGAGUUUGAGAACUACAAAAACCAUCGACCGUUGACUGUACCUUGAGGUUUAUCACAAACCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038550
          miRBase ID: cli-miR-181a-5p
          Mature miRNA Sequence: AACAUUCAACGCUGUCGGUGAGUU
          Chromosome Location: NA

          Ophiophagus hannah

          Stem-loop ID oha-mir-181a-1
          Stem-loop Accession # MI0031375
          Stem-loop Sequence
          CUUACGAUAGCUUCAGCGAACAUUCAACGCUGUCGGUGAGUUUGGUCCCUCAAGAGAAACCACCGAUCGUUGACUGUACCUUGAGGUUUAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036757
          miRBase ID: oha-miR-181a-5p
          Mature miRNA Sequence: AACAUUCAACGCUGUCGGUGAGUU
          Chromosome Location: NA

          Stem-loop ID oha-mir-181a-2
          Stem-loop Accession # MI0031376
          Stem-loop Sequence
          GGUAAAAGUUCUCGGGGAACAUUCAACGCUGUCGGUGAGUUUUGUCAAUGAGGUCAAACCAUCGACUGUUGAGUGUACCCUGCGCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036757
          miRBase ID: oha-miR-181a-5p
          Mature miRNA Sequence: AACAUUCAACGCUGUCGGUGAGUU
          Chromosome Location: NA

          Pig

          Stem-loop ID ssc-mir-181a-1
          Stem-loop Accession # MI0010686
          Stem-loop Sequence
          GAGUUUAGAGGUUGCUUCAGUGAACAUUCAACGCUGUCGGUGAGUUUGGAAUUCAAAUCAAAACCAUCGACCGUUGAUUGUACCCUAUGGCCAACCAUCAGCUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0010191
          miRBase ID: ssc-miR-181a
          Mature miRNA Sequence: AACAUUCAACGCUGUCGGUGAGUU
          Chromosome Location: NA

          Stem-loop ID ssc-mir-181a-2
          Stem-loop Accession # MI0013106
          Stem-loop Sequence
          ACUCCAAGGAACAUUCAACGCUGUCGGUGAGUUUGGGAUUUGAAAAAACCACCGACCGUUGACUGUACCUUGGGGUUCUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0010191
          miRBase ID: ssc-miR-181a
          Mature miRNA Sequence: AACAUUCAACGCUGUCGGUGAGUU
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ MicroRNA Assay : 005861_mat
          mirVana® miRNA inhibitor : MH11451

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline