Search
The QuantStudio 6 and 7 Flex real-time PCR systems are designed for versatility. Easily interchange between 96-well (0.1 and 0.2 mL) and 384-well formats. The systems can detect differences as small as 1.5-fold in singleplex reactions with 10 logs dynamic range. To help you to quickly adapt to changing requirements and protect your investment, the QuantStudio 6 system can be upgraded to a QuantStudio 7 system at any time.
The QuantStudio 7 Flex system can also run a TaqMan Array card (a 384-well card that enables the loading of 384 targets with 8 pipetting steps in less than 10 minutes).
In addition to the common features of all QuantStudio systems, the QuantStudio 6 and 7 Flex systems offer:
Easily change the block from the front in less than a minute with no tools required
The QuantStudio 6 Flex system can be seamlessly upgraded to a QuantStudio 7 Flex system by a field service engineer, adding automation compatibility, and the workflow efficiencies of TaqMan Array cards.
The intuitive software, easy touch-screen setup, and block changes are designed to let you get started right away.
Features that are unique to the QuantStudio 7 Flex system:
Interchangeable 96-well (0.1 and 0.2 mL), 384-well, plus TaqMan Array card blocks
Run hundreds of real-time PCR reactions effortlessly using the Thermo Scientific Orbitor RS2 Microplate Mover. The QuantStudio 7 Flex system can maximize your automated throughput capabilities.
Features six decoupled excitation and emission filter channels with 21 filter combinations for maximum multiplexing and chemistry flexibility.
Maximize the performance of your QuantStudio 6 and 7 Flex systems with the following calibration kits.
| Category | Specification | QuantStudio 6 Flex system | QuantStudio 7 Flex system |
|---|---|---|---|
| Flexible format |
Interchangeable blocks |
||
| Efficiency and productivity |
Small footprint | ||
| Multiplexing |
5 |
6 (21 filter combinations) |
|
| Remote monitoring | |||
| Intuitive touchscreen | |||
| Automation compatible |
|||
| Features to assist with 21 CFR part 11 compliance |
* |
* |
|
| Dimensions |
112.5 x 90.7 x 74.2 cm (H x W x D) | 112.5 x 90.7 x 74.2 cm (H x W x D) | |
| Maximize uptime |
Smart Help |
||
| Minimal calibration for easy maintenance |
|||
| Support |
On-site installation by professionally trained field service engineers | ||
| On-site training package conducted by field applications scientists | |||
| Augmented reality and remote desktop support capability |
*Available as an option.