Product Image
Applied Biosystems™
MeltDoctor™ HRM Calibration Plate, 384-well
Applied Biosystem® ready-to-use MeltDoctor™ HRM Calibration Plates contain all the components required for pure dye and HRM calibration, reducing the자세히 알아보기
Have Questions?
카탈로그 번호수량
44255591 x 384-well plate
카탈로그 번호 4425559
제품 가격(KRW)
1,120,000
온라인 행사
Ends: 30-Jun-2026
1,244,000
할인액 124,000 (10%)
Each
수량:
1 x 384-well plate
제품 가격(KRW)
1,120,000
온라인 행사
Ends: 30-Jun-2026
1,244,000
할인액 124,000 (10%)
Each
Applied Biosystem® ready-to-use MeltDoctor™ HRM Calibration Plates contain all the components required for pure dye and HRM calibration, reducing the complexity of getting started with HRM experimentation. These plates provide thermal and optical calibration required for accurate and reproducible discrimination between melt curves.
For Research Use Only. Not for use in diagnostic procedures.
사양
보정된 염료HRM (High Resolution Melting) Dye
보정 유형Thermal Calibration, Optic Calibration
용도(애플리케이션)Real Time PCR (qPCR)
용도(장비)7900HT Fast System
형식384-well Plate
플레이트 호환성Calibration Plate
제품라인MeltDoctor
수량1 x 384-well plate
배송 조건Dry Ice
제품 유형HRM Calibration Plate
Unit SizeEach
구성 및 보관
1 plate, 384-well format
Guaranteed minimum shelf life is 60 days (exact expiry date printed on product and CofA).
자주 묻는 질문(FAQ)
How long should the amplicon be for HRM analysis?
We recommend to use 50-250 bp long PCR amplicons. Typically, shorter amplicons can distinguish the genotypes for a SNP better, especially for Type III and Type IV SNPs. This is simply because a single base variation affects the melting behavior of a 100 bp amplicon stronger than of a 500 bp amplicon, for example. In longer fragments, the risk of covering multiple mutations is also increased.
What can be used as positive control reactions in HRM analysis?
Positive control reactions should contain template DNA with a known sequence. In SNP genotyping experiments, this could be a sample with a known genotype. Positive control(s) for all genotypes should be included where possible to serve as a reference in melting curve comparison and assigning genotypes for test samples. In mutation scanning experiments, a sample with a wild type sequence could serve as a positive control. The controls should preferably have the same DNA concentration as their corresponding test samples. Control DNA should also be eluted and/or diluted in the same buffer as the samples.
In HRM analysis, when is it better to use a 3-step cycling protocol rather than a 2-step protocol?
A 3-step cycling protocol is recommended for the analysis of complicated (especially Type IV SNP) targets, amplicons longer than 200 bp, and amplicons with a primer annealing temperature that is less than 60 degrees C.
The HRM software will not accept my calibration file. What is wrong?
Check that there are no outliers on the plate. You cannot omit any wells on the HRM calibration plate.
I have a 7900HT Fast Real-Time PCR System, and cannot open the data file in HRM software. What could be causing this?
There are a few possibilities. First, make sure the HRM Software version is v2.0.1, and the 7900HT Fast Real-Time PCR System software version is v2.3 or above. Second, check that the run method used was as recommended in the HRM protocol; make sure the ramp rate for the dissociation stage is 1%. Then try to open the calibration file from the HRM Software; if it does not open, the calibration file is defective. The defects could be due to a bad calibration plate or instrument uniformity issue.