Introduction
Cell and nuclear extraction are vital techniques to isolate cellular components for analysis. These methods are applicable to both adherent and suspension cells. Adherent cells are scraped off their growth surface, while suspension cells are collected by centrifugation. Both types of cells undergo lysis and centrifugation to yield cytoplasmic and nuclear fractions. These high-quality extracts are essential for various research applications, including protein quantification and signal transduction studies.
This protocol has been developed for rapid isolation of cytokines and signaling molecules, using a non-abrasive extraction reagent for total protein extraction. The method is sensitive and allows for the detection of disease-associated biomarker fluctuations. The prepared extracts are compatible with a variety of immunoassays, including ELISA and the Luminex platform, as well as conventional protein assays. Typically, this extraction method is cost-effective, versatile, and can be completed in less than 15 minutes.
Search available antibody pairs and ELISA kits  Browse available ELISA reagents
Materials
Note: 1X Cell extraction buffer may be divided into 1X aliquots in microcentrifuge tubes and stored at –20°C until ready for use. Thaw on ice prior to extracting cells.
Additional recommended materials
Protocol tips
Addition of the protease inhibitor cocktail and PMSF is necessary to inhibit proteolysis in cell extracts. Just prior to use, supplement the cell extraction buffer with 1 mM PMSF and protease inhibitor to make complete cell extraction buffer. Prepare 0.3 M PMSF stock in DMSO and add final concentration of 1 mM PMSF (i.e., 17 μL of 0.3M PMSF stock in 5 mL cell extraction buffer). PMSF is very unstable and must be added just before use, even if it was added previously. For Protease Inhibitor addition, we recommend using Halt Protease Inhibitor Cocktail, reconstituted to 1X (10 μL of 100X Halt Protease Inhibitor Cocktail in 1 mL of cell extraction buffer). The stability of protease inhibitor-supplemented cell extraction buffer is 24 hours at 4°C.
Procedure
Processing cell culture samples
  1. Following the end of the desired cell culture time (adherent or suspension cells), pipette the medium into a microcentrifuge tube, and immediately put on ice.
  1. Centrifuge at 1,400 rpm for 1 minute at 4°C.
  1. Collect the cell culture supernatant into a clean microcentrifuge tube.
  1. Perform the ELISA immediately or store at –80°C until ready for use.
Processing cell lysates
Note: This method can be used to produce relatively large quantities of cell extracts with each of the stimulation regimes studied. The wash steps included in this procedure help to minimize medium components in the cell extracts.
  1. Estimate cell density. For suspension cells, enumerate cell density by counting in a hemacytometer. For adherent cells, estimate the cell density by visual inspection under a microscope.
    Note
    :
    Confluence levels of 70–80% are found to be optimal for many signal transduction studies.
  1. Stimulate the cells as desired.
  1. Transfer the cells into clean 15 mL conical tubes. Suspension cells—aliquot the desired number of cells in medium into clean 15 mL conical tubes. Adherent cells—remove the cells from their vessel by scraping. Transfer the medium containing the detached cells into clean 15 mL conical tubes.
  1. Collect the cells by centrifugation at 300 x g for 7 minutes at 4°C.
  1. Aspirate the medium.
  1. Resuspend the pellet in ice-cold 1X Phosphate-buffered Saline (PBS).
  1. Collect the cells by centrifugation at 300 x g for 7 minutes at 4°C.
  1. Aspirate the PBS.
  1. Lyse the cells by pipetting complete cell extraction buffer into each tube. It is recommended to use 1 mL of complete cell extraction buffer per 108 cells. It is important to note that this value may require optimization for each specific application.
  1. Transfer the lysates to clean microcentrifuge tubes.
  1. Vortex the mixture, then incubate the mixture on ice for 30 minutes, with occasional vortexing.
  1. Clarify the lysates by centrifugation at 14,000 rpm (13,000 x g) for 10 minutes at 4°C.
  1. Transfer the clarified cell extracts into clean microcentrifuge tubes.
  1. The clarified cell extracts should be stored at –80°C until ready for analysis. Avoid repeated freeze-thaw cycles. In preparation for performing the assay, allow the samples to thaw on ice. Mix well prior to analysis.
  1. Determine protein concentration using protein assay of your choice, or ready-to-use Quant-iT Reagents and Kits for 20-2,000 Samples, such as the Quant-iT Protein Assay Kit. Cell extracts prepared by this method routinely have a protein concentration between 1 and 10 mg/mL.
  1. Certain analytes require a sample treatment step. Please refer to the analyte specific protocol for details on sample treatment recommendations.
  1. All cell extracts require dilution by a factor of at least 1:10 in Standard Diluent Buffer (included in the kit) before analysis with Invitrogen ELISA kits.
Materials
Note:1X Cell extraction buffer may be divided into 1X aliquots in microcentrifuge tubes and stored at –20°C until ready for use. Thaw on ice prior to extracting cells.
Additional recommended materials
Protocol tips
To inhibit proteolysis in cell extracts, it is essential to add a protease inhibitor cocktail and PMSF. Just before use, supplement the cell extraction buffer with 1 mM PMSF and Protease Inhibitor to prepare complete cell extraction buffer. Prepare 0.3 M PMSF stock in DMSO and add it to achieve a final concentration of 1 mM PMSF (i.e., 17 μL of 0.3 M PMSF stock in 5 mL cell extraction buffer). PMSF is very unstable and must be added immediately before use, regardless of prior addition. For the Protease Inhibitor, we recommend using Halt Protease Inhibitor Cocktail, reconstituted to 1X (10 μL of 100X Halt Protease Inhibitor Cocktail in 1 mL of cell extraction buffer). The stability of cell extraction buffer supplemented with protease inhibitor is 24 hours at 4°C.
Procedure
Note: This protocol has been successfully applied to several cell lines of human origin. Researchers should optimize the nuclear extraction procedure for their own applications.
  1. Collect cells (5 x 106) in Phosphate-buffered Saline (PBS) by centrifugation (suspension cells) or by cell scraping or use a cell dissociation reagent (adherent cells).
  1. Wash the cells twice with cold PBS.
  1. Remove and discard the supernatant and collect the cell pellet. 
  1. Transfer the cells into a prechilled microcentrifuge tube.
  1. Gently resuspend cells in 500 μL of 1X hypotonic buffer by pipetting up and down several times. Incubate on ice for 15 minutes.
  1. Add 25 μL of 10% NP-40 detergent and vortex for 10 seconds at highest setting.
  1. Centrifuge the homogenate for 10 minutes at 3,000 rpm at 4°C.
  1. Transfer and save the supernatant. This supernatant contains the cytoplasmic fraction. The pellet is the nuclear fraction.
  1. Resuspend nuclear pellet in 50 μL of complete cell extraction buffer for 30 minutes on ice with vortexing at 10-minute intervals.
  1. Centrifuge for 30 minutes at 14,000 x g at 4°C. Transfer the supernatant (nuclear fraction) into a clean microcentrifuge tube.
  1. Aliquot and store at –80°C. The nuclear extracts are ready for assay.
  1. Quantitate protein concentration using protein assay of your choice, or ready-to-use Quant-iT Reagents and Kits for 20–2,000 Samples, such as the Quant-iT Protein Assay Kit.
Catalog # Name Size Price (USD) Qty
FNN0011 Cell Extraction Buffer Each
185.65
Online Exclusive
193.00
Save 7.35 (4%)
69715 Pierce™ Microcentrifuge Tubes, 1.5 mL Each
59.65
Online Exclusive
64.00
Save 4.35 (7%)
339651 15 mL Conical Centrifuge Tube, racked, Pack of 25 Case of 500
369.65
Online Exclusive
406.00
Save 36.35 (9%)
339650 15 mL Conical Centrifuge Tube, bulk, Pack of 50 Case of 500
357.65
Online Exclusive
392.00
Save 34.35 (9%)
78429 Halt™ Protease Inhibitor Cocktail (100X), 5 mL Each
455.65
Online Exclusive
506.00
Save 50.35 (10%)
78430 Halt™ Protease Inhibitor Cocktail (100X), 24 x 100 μL Each
236.65
Online Exclusive
263.00
Save 26.35 (10%)
78438 Halt™ Protease Inhibitor Cocktail (100X), 10 mL Each
646.65
Online Exclusive
730.00
Save 83.35 (11%)
87786 Halt™ Protease Inhibitor Cocktail (100X), 1 mL Each
127.65
Online Exclusive
141.00
Save 13.35 (9%)
36978 PMSF Protease Inhibitor Each
105.65
Online Exclusive
117.00
Save 11.35 (10%)
D12345 DMSO, Anhydrous Each
140.00
28348 Pierce™ 20X Phosphate Buffered Saline, 500 mL Each
139.65
Online Exclusive
152.00
Save 12.35 (8%)
Q33210 Quant-iT™ Protein Assay Kit Each
609.65
Online Exclusive
654.00
Save 44.35 (7%)
85124 NP-40 Surfact-Amps™ Detergent Solution, 50 mL Each
163.65
Online Exclusive
196.00
Save 32.35 (17%)
28324 NP-40 Surfact-Amps™ Detergent Solution, 6 x 10 mL Each
151.65
Online Exclusive
181.00
Save 29.35 (16%)
85125 NP-40 Surfact-Amps™ Detergent Solution, 500 mL Each
963.65
Online Exclusive
1,154.00
Save 190.35 (16%)
20012027 PBS, pH 7.2, 1 x 500 mL, No Sodium Pyruvate Each
31.65
Online Exclusive
33.21
Save 1.56 (5%)
20012050 PBS, pH 7.2, 10 x 500 mL, No Sodium Pyruvate Each
268.65
Online Exclusive
286.00
Save 17.35 (6%)
20012043 PBS, pH 7.2, 1 x 1000 mL, No Sodium Pyruvate Each
47.65
Online Exclusive
53.00
Save 5.35 (10%)
Microplate readers
Compare now
Stylesheet for Classic Wide Template adjustments
For Research Use Only. Not for use in diagnostic procedures.