Halt™ Protease Inhibitor Cocktail (100X)
Thermo Scientific™

Halt™ Protease Inhibitor Cocktail (100X)

Thermo Scientific Halt Protease Inhibitor Cocktail (100X) are ready-to-use concentrated stock solutions of protease inhibitors for addition to samples toRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
784295 mL
7843024 x 100 μL
7843810 mL
877861 mL
Catalog number 78429
Price (USD)
450.65
Online exclusive
518.00
Save 67.35
Each
Quantity:
5 mL
Request bulk or custom format
Price (USD)
450.65
Online exclusive
518.00
Save 67.35
Each

Thermo Scientific Halt Protease Inhibitor Cocktail (100X) are ready-to-use concentrated stock solutions of protease inhibitors for addition to samples to prevent proteolytic degradation during cell lysis and protein extraction.

Features of Halt Protease Inhibitor Cocktail, (100X):

  • Convenient—the refrigerator-stable, 100X liquid format is more effective and easier to use than individual inhibitors or other formats; just pipette exactly the amount you need
  • No proprietary ingredients—fully disclosed formulation contains optimized concentrations of six broad-spectrum protease inhibitors AEBSF, aprotinin, bestatin, E-64, leupeptin and pepstatin A stabilized in high-quality dimethylsulfoxide (DMSO)
  • Multiple package sizes—choose among several package sizes, including single-use microtubes that keep protease inhibitors fresh and reduce the risk of reagent contamination
  • Regular formulation—includes a separate vial of EDTA solution for optional metalloprotease inhibition
  • Lysis buffer compatible—use with Thermo Scientific Pierce Cell Lysis Buffers or nearly any other commercial or homemade detergent-based lysis reagent. Add 10 μL of concentrated cocktail per 1 mL of lysis buffer to ensure complete protection of the resulting protein extract.

Applications:

  • Protection of intact, active cellular proteins from degradation by endogenous proteases
  • Inhibition of protease activity from lysates produced by a variety of methods including Thermo Scientific Pierce Cell Lysis Reagents, other commercially formulated cell or tissue lysis products, sonication, French press, blender homogenization or repetitive freeze/thaw cycling
  • Screening of extracts for proteolytic activity
  • The study of proteolysis in the regulation of cellular processes

Proteases make up a large category of enzymes, including endopeptidases and exopeptidases, which catalyze the hydrolytic breakdown of proteins into peptides or amino acids in a wide variety of cell and tissue types. Proteases are regulated and compartmentalized in living cells, but cell lysis releases and mixes them with other extracted proteins. Their activity can thus lead to the loss of a large number of valuable proteins, adversely affecting downstream applications. For this reason, it is common practice to spike cell lysis buffers or cell extracts with a cocktails of compounds known to inactivate or inhibit proteases. Halt Protease Inhibitor Cocktails effectively inhibit serine-proteases, cysteine-proteases, aspartic acid-proteases and aminopeptidases that are typically present in cellular lysate samples. Regular and EDTA-free formulations provide for inhibition of metalloproteases and compatibility with 2D electrophoresis, respectively.

Related Products

Halt™ Protease Inhibitor Single-Use Cocktail, EDTA-free (100X) (Cat. No. 78425)

For Research Use Only. Not for use in diagnostic procedures.
Specifications
BufferLysis Buffer
InhibitorsProtease Inhibitor
Quantity5 mL
Reagent TypeProtease Inhibitor
FormLiquid
Product LineHalt
Product TypeProtease Inhibitor Cocktail
Unit SizeEach
Contents & Storage
Upon receipt store at 4°C

Frequently asked questions (FAQs)

What are the compositions of the Halt Protease Inhibitor Cocktail, EDTA-Free (100X) (Cat. No. 87785) and Halt Protease Inhibitor Cocktail (100X) (Cat. No. 87786)?

Both formulations are the same and have the following composition:

What is the shelf life of Halt Protease Inhibitor Cocktail (100X)?

When stored as recommended at 4 degrees C, Halt Protease Inhibitor Cocktail (100X) is guaranteed stable for 1 year from the date of receipt.

Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.

Can I use just part of the Pierce Protease Inhibitor XL Capsules, EDTA-Free and save part of the capsule for later use?

We do not recommend “splitting” the capsule into smaller portions. Researchers using less than 500 mL should choose a different format (liquid or tablet) of protease inhibitors.

Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.

What formats of protease inhibitors do you offer?

We offer standalone protease inhibitors (AEBSF, Aprotinin, PMSF, EDTA, etc.), as well as cocktails or combinations of protease inhibitors in liquid, tablet, and larger capsule formats. These cocktails of protease inhibitors can also be purchased in combination with a cocktail of phosphatase inhibitors, in both liquid and tablet form, for even greater convenience.

Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.

What protease inhibtors are present in the Halt Protease Inhibitor Single-Use Cocktail?

The inhibitors included in the Halt Protease Inhibitor Single-Use Cocktail (Cat. No. 78430) are: AEBSF, Aprotinin, Bestatin, E64, Leupeptin, Pepstatin A, and EDTA.

Citations & References (4)

Citations & References
Abstract
Overexpression of TBX3 suppresses tumorigenesis in experimental and human cholangiocarcinoma.
Authors:Deng S,Lu X,Wang X,Liang B,Xu H,Yang D,Cui G,Yonemura A,Paine H,Zhou Y,Zhang Y,Simile MM,Urigo F,Evert M,Calvisi DF,Green BL,Chen X
Journal:Cell death & disease
PubMed ID:38909034
TBX3 behaves as a tumor suppressor or oncoprotein across cancer. However, TBX3 function remains undetermined in intrahepatic cholangiocarcinoma (iCCA), a deadly primary liver malignancy with few systemic treatment options. This study sought to investigate the impact of TBX3 on iCCA. We found that overexpression of TBX3 strongly inhibited human iCCA ... More
Diverse mechanisms of DDX3Y suppression by DDX3X.
Authors:Xu X,Wei S
Journal:bioRxiv : the preprint server for biology
PubMed ID:39975375
The DEAD-box RNA helicase DDX3X has important roles in development and disease. Loss of DDX3X during developmental and pathological processes such as tumorigenesis can lead to compensatory upregulation of the close paralog DDX3Y in males, which may underlie the sexual dimorphism displayed by some DDX3X-associated diseases. However, how DDX3X cross-regulates ... More
FOXO1 modulates osteoblast differentiation.
Authors:Siqueira MF,Flowers S,Bhattacharya R,Faibish D,Behl Y,Kotton DN,Gerstenfeld L,Moran E,Graves DT
Journal:Bone
PubMed ID:21281751
Forkhead box O1 (FOXO1) is upregulated during bone formation and in response to stimulation by bone morphogenetic proteins. Studies presented here examined the functional role of FOXO1 in a well defined culture system in which pre-osteoblastic cells undergo terminal differentiation in vitro. Mineralizing cultures of MC3T3-E1 cells were examined with ... More
Glucagon-like peptide 1 inhibits cell apoptosis and improves glucose responsiveness of freshly isolated human islets.
Authors:Farilla L, Bulotta A, Hirshberg B, Li Calzi S, Khoury N, Noushmehr H, Bertolotto C, Di Mario U, Harlan DM, Perfetti R
Journal:Endocrinology
PubMed ID:12960095
The peptide hormone, glucagon-like peptide 1 (GLP-1), has been shown to increase glucose-dependent insulin secretion, enhance insulin gene transcription, expand islet cell mass, and inhibit beta-cell apoptosis in animal models of diabetes. The aim of the present study was to evaluate whether GLP-1 could improve function and inhibit apoptosis in ... More