Halt™ Protease Inhibitor Cocktail, EDTA-Free (100X)
Thermo Scientific™

Halt™ Protease Inhibitor Cocktail, EDTA-Free (100X)

Inhibit proteolytic degradation during cell lysis and protein extraction with these ready-to-use concentrated stock solutions of protease inhibitors.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
7842524 x 100 μL
784375 mL
7843910 mL
877851 mL
Catalog number 78425
Price (USD)
221.65
Online exclusive
252.00
Save 30.35
Each
Quantity:
24 x 100 μL
Request bulk or custom format
Price (USD)
221.65
Online exclusive
252.00
Save 30.35
Each

Thermo Scientific Halt Protease Inhibitor Cocktail, EDTA-free (100X) are ready-to-use concentrated stock solutions of protease inhibitors for addition to samples to prevent proteolytic degradation during cell lysis and protein extraction.

Features of Halt Protease Inhibitor Cocktail, EDTA-free (100X):

  • Convenient—the refrigerator-stable, 100X liquid format is more effective and easier to use than individual inhibitors or other formats; just pipette exactly the amount you need
  • No proprietary ingredients—fully disclosed formulation contains optimized concentrations of six broad-spectrum protease inhibitors AEBSF, aprotinin, bestatin, E-64, leupeptin and pepstatin A stabilized in high-quality dimethylsulfoxide (DMSO)
  • Multiple package sizes—choose among several package sizes, including single-use microtubes that keep protease inhibitors fresh and reduce the risk of reagent contamination
  • EDTA-free formulations—ensures compatibility with isoelectric focusing
  • Lysis buffer compatible—use with Thermo Scientific Pierce Cell Lysis Buffers or nearly any other commercial or homemade detergent-based lysis reagent. Add 10 μL of concentrated cocktail per 1 mL of lysis buffer to ensure complete protection of the resulting protein extract.

Applications:

  • Protection of intact, active cellular proteins from degradation by endogenous proteases
  • Inhibition of protease activity from lysates produced by a variety of methods including Thermo Scientific Pierce Cell Lysis Reagents, other commercially formulated cell or tissue lysis products, sonication, French press, blender homogenization or repetitive freeze/thaw cycling
  • Screening of extracts for proteolytic activity
  • The study of proteolysis in the regulation of cellular processes

Proteases make up a large category of enzymes, including endopeptidases and exopeptidases, which catalyze the hydrolytic breakdown of proteins into peptides or amino acids in a wide variety of cell and tissue types. Proteases are regulated and compartmentalized in living cells, but cell lysis releases and mixes them with other extracted proteins. Their activity can thus lead to the loss of a large number of valuable proteins, adversely affecting downstream applications. For this reason, it is common practice to spike cell lysis buffers or cell extracts with a cocktails of compounds known to inactivate or inhibit proteases. Halt Protease Inhibitor Cocktails effectively inhibit serine-proteases, cysteine-proteases, aspartic acid-proteases and aminopeptidases that are typically present in cellular lysate samples. Regular and EDTA-free formulations provide for inhibition of metalloproteases and compatibility with 2D electrophoresis, respectively.

Related Products

  • Halt™ Protease Inhibitor Single-Use Cocktail (100X) (Cat. No. 78430)
For Research Use Only. Not for use in diagnostic procedures.
Specifications
BufferLysis Buffer
InhibitorsProtease Inhibitor
Quantity24 x 100 μL
Reagent TypeProtease Inhibitor
FormLiquid
Product LineHalt
Product TypeProtease Inhibitor Cocktail
Unit SizeEach
Contents & Storage
Upon receipt store at 4°C

Frequently asked questions (FAQs)

What are the compositions of the Halt Protease Inhibitor Cocktail, EDTA-Free (100X) (Cat. No. 87785) and Halt Protease Inhibitor Cocktail (100X) (Cat. No. 87786)?

Both formulations are the same and have the following composition:

What is the shelf life of Halt Protease Inhibitor Cocktail, EDTA-free (100X) (Cat. No. 78425)?

The Halt Protease Inhibitor Cocktail, EDTA-free (100X) (Cat. No. 78425) is covered under our general 1-year warranty and is guaranteed to be fully functional for 12 months from the date of shipment, if stored as recommended. Please see section 8.1 of our Terms & Conditions of Sale for more details.

Can I use just part of the Pierce Protease Inhibitor XL Capsules, EDTA-Free and save part of the capsule for later use?

We do not recommend “splitting” the capsule into smaller portions. Researchers using less than 500 mL should choose a different format (liquid or tablet) of protease inhibitors.

Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.

What formats of protease inhibitors do you offer?

We offer standalone protease inhibitors (AEBSF, Aprotinin, PMSF, EDTA, etc.), as well as cocktails or combinations of protease inhibitors in liquid, tablet, and larger capsule formats. These cocktails of protease inhibitors can also be purchased in combination with a cocktail of phosphatase inhibitors, in both liquid and tablet form, for even greater convenience.

Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.

Citations & References (2)

Citations & References
Abstract
miR-34c is downregulated in prostate cancer and exerts tumor suppressive functions.
Authors:Hagman Z, Larne O, Edsjö A, Bjartell A, Ehrnström RA, Ulmert D, Lilja H, Ceder Y
Journal:Int J Cancer
PubMed ID:21351256
MicroRNAs (miRNAs) are small noncoding RNAs that post-transcriptionally regulate gene expression. There have been several reports of miRNA deregulation in prostate cancer (PCa) and the biological evidence for an involvement of miRNAs in prostate tumorigenesis is increasing. In this study, we show that miR-34c is downregulated in PCa (p = ... More
Glucagon-like peptide 1 inhibits cell apoptosis and improves glucose responsiveness of freshly isolated human islets.
Authors:Farilla L, Bulotta A, Hirshberg B, Li Calzi S, Khoury N, Noushmehr H, Bertolotto C, Di Mario U, Harlan DM, Perfetti R
Journal:Endocrinology
PubMed ID:12960095
The peptide hormone, glucagon-like peptide 1 (GLP-1), has been shown to increase glucose-dependent insulin secretion, enhance insulin gene transcription, expand islet cell mass, and inhibit beta-cell apoptosis in animal models of diabetes. The aim of the present study was to evaluate whether GLP-1 could improve function and inhibit apoptosis in ... More