PMSF Protease Inhibitor
Thermo Scientific™

PMSF Protease Inhibitor

Thermo Scientific PMSF is a protease inhibitor that reacts with serine residues to inhibit trypsin, chymotrypsin, thrombin and papain.Features ofRead more
Have Questions?
Catalog NumberQuantity
369785 g
Catalog number 36978
Price (USD)
107.65
Online exclusive
120.00
Save 12.35
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
107.65
Online exclusive
120.00
Save 12.35
Each
Thermo Scientific PMSF is a protease inhibitor that reacts with serine residues to inhibit trypsin, chymotrypsin, thrombin and papain.

Features of PMSF:

• Targets serine proteases, including trypsin, chymotrypsin, thrombin and papain
• Effective in most protein solutions at 0.1 to 1 mM final concentration
• Because PMSF has limited water solubility, it must be dissolved in a small amount of solvent, such as ethanol, methanol or isopropyl alcohol before addition to a buffer

PMSF is an abbreviation for phenylmethylsulfonyl fluoride, the most common chemical name for this small compound. In addition to inhibiting serine proteases, PMSF will also inhibit cysteine proteases, like papain (reversible by DTT treatment), and mammalian acetylcholinesterase.

Properties of PMSF:
• Synonyms: alpha-Toluenesulfonyl fluoride; Benzylsulfonyl fluoride; Phenylmethylsulfonyl fluoride
• Formula: C6H5CH2SO2F
• Molecular weight: 174.19
• CAS Number: 329-98-6; EC Number: 206-350-2
• R/S Codes: R 25-34; S 26-36/37/39-45
• Storage conditions: room temperature in a dry place protected from light
• Working solution: dissolve PMSF in isopropanol at 1.74 mg/mL (10 mM); store in aliquots at -20°C.

Specifications of PMSF (Part No. 36978):
• Appearance: white powder
• Purity: > 99.0 %
• Melting point: 90-95°C
• Humidity: < 0.1 %

Related Products
Pierce™ Protease and Phosphatase Inhibitor Mini Tablets, EDTA Free
Pierce™ Protease Inhibitor Mini Tablets, EDTA Free
For Research Use Only. Not for use in diagnostic procedures.
Specifications
DescriptionPierce PMSF Protease Inhibitor
InhibitorsProtease Inhibitor
Quantity5 g
Reagent TypeProtease Inhibitor
FormPowder
Product TypeProtease Inhibitor
Unit SizeEach
Contents & Storage
Store in original container protected from direct sunlight in a dry, cool and well-ventilated area, between the following temperatures: 20°C to 25°C.

Frequently asked questions (FAQs)

How should I reconstitute and store PMSF Protease Inhibitor (Cat. No. 36978)?

We recommend dissolving PMSF Protease Inhibitor (Cat. No. 36978) in isopropanol at 1.74 mg/mL (10 mM) and storing in aliquots at -20 degrees C.

Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.