Recent searches Clear History
聯絡我們
ImmunoCAP 檢測解讀指南
有了檢測解讀指南的支持,您能夠安心診斷和管理過敏。
利用有關如何解讀 ImmunoCAPTM 特異性 IgE 檢測結果及潛在臨床意義的簡要資訊,協助您的診斷和患者管理計畫。
如何解讀 ImmunoCAP 特定 IgE 檢測結果
結果顯示患者對特定過敏原的敏感程度,表示 ImmunoCAP 特異性 IgE 檢測可協助診斷臨床過敏。 
Results are reported in the amount of IgE antibodies for a specific allergen in kilounits of allergen per liter of blood (kUA/l). 
Results ≥0.1 kUA/l indicate an allergen sensitization, which can be used to help clinically diagnose allergy as long as patient history is taken into account. ImmunoCAP tests alone cannot diagnose a clinical allergy.1
Sensitization levels vary from patient to patient, and there is no standardized numerical amount which indicates the severity of a reaction. Therefore, each patient’s personal symptom history must be considered before diagnosing.
Example:
  • Patient A has a cat allergen sensitization of 10 kUA/l but does not react to cats.
    Patient A is sensitized to cats but not allergic.
  • Patient B has a cat allergen sensitization of 5 kUA/l and experiences severe rhinitis when exposed.
    Patient B may be clinically diagnosed with a cat allergy. 
過敏原成分在檢測中的角色
深入瞭解過敏,進而改善診斷結果。 
過敏原成分檢測會測量對完整過敏原中特定蛋白質和/或碳水化合物的過敏性敏感作用,讓您更深入瞭解交叉反應性、潛在嚴重度和潛在耐受性。2
使用過敏原成分進行檢測可得到最佳的:2,3
  • 過敏診斷
  • 過敏管理
  • 深入分析特定的治療選項 
呼吸道過敏結果解讀指南
地區性呼吸道套組針對患者居住地區特有的環境過敏原,提供更加個人化的檢測與管理。 
Pet Allergy Interpretation Guide
過敏原成分
透過寵物過敏的過敏原成分解讀,可深入瞭解交叉反應性、氣喘風險,以及患者是對公犬或母犬過敏。3
探索地區特定呼吸道過敏套組的力量。 
食物過敏結果解讀指南
利用主要食物過敏原、食品過敏原成分指南,提升食物過敏患者的管理。 
Egg Allergy Interpretation Guide
Food Allergy Interpretation Guide
Milk Allergy Interpretation Guide
Sesame Allergy Interpretation Guide
Tree Nut Allergy Interpretation Guide
Wheat Interpretation Guide
Peanut Allergy Interpretation Guide
過敏原成分
透過食物 (例如堅果、牛奶、蛋等等) 過敏原成分解讀指南,可深入瞭解交叉反應性、潛在嚴重度和烘焙/原型食物耐受性。3
探索 ImmunoCAP 特異性 IgE 檢測如何幫助診斷食物過敏。
探索專為協助診斷和患者管理工作而設計的全套 ImmunoCAP 特異性 IgE 檢測解讀指南。 
參考資料
  1. ImmunoCAP Specific IgE Directions for Use 2020 for the Phadia Laboratory Systems.
  2. Canonica, G.W., et al., A WAO - ARIA - GA(2)LEN consensus document on molecular-based allergy diagnostics.World Allergy Organ J, 2013.6(1): p. 17. 
  3. Matricardi, P.M., et al., EAACI Molecular Allergology User's Guide.Pediatr Allergy Immunol, 2016.27 Suppl 23: p 77, 79-81, 97, 100, 142-143, 152, 158