These products may not be cleared for use in your country. Please contact your sales representative for information about specific product availability.
ImmunoCAP™ Total IgE measures the total amount of circulating IgE in human serum or plasma samples.1 IgE antibodies can appear as a result of sensitization to allergens. In many patients suffering from asthma, rhinitis, or atopic dermatitis, the concentration of total IgE may be elevated.1,3-4
ImmunoCAP Total IgE is a fully quantitative test that provides accurate levels of circulating IgE within a wide measuring range between 2 to 5000 kU/L.1 For patients with low serum IgE levels, ImmunoCAP Total IgE Low Range offers quantitative measurement between 0.04-100 kU/L.2