Method for selecting negative cell isolation of untouched cells
Negative cell isolation is a process targeting unwanted cell types for removal, leaving the desired cells untouched. The method involves using Dynabeads magnetic beads with ligands to target surface proteins of specific cells. In contrast, positive cell isolation captures the desired cells using Dynabeads paired with ligands to target the specific cell surface protein. You can read more about Dynabeads and their expanding applications.
Negative cell isolation with Dynabeads is a gentle, tube-based method. Your cells are not exposed to the stress of being passed through a column. The isolated pure, viable, and truly untouched target cells can be directly analyzed in a flow cytometer and used in any functional assay.
You are guaranteed:
  • Minimal stress on cells  - no interaction during isolation, target cells are left bead- and antibody-free
  • No columns - the cells are not exposed to the stress of being passed through a dense column
  • High purity, viability and recovery - negatively isolated cells are in perfect shape for any applifcation
Performance of negative cell isolation showing purity of monocytes among PBMC before and after isolation.
The performance of negative cell isolation with Dynabeads® is exemplified by these figures. The top panels show the purity of monocytes among PBMC prior to (20%) and after isolation (94%) using Dynabeads® Untouched™ Human Monocytes. The below panels show how isolated monocytes differentiate into dendritic cells after 5 days in culture (d5) with IL-4 and GM-CSF as shown by increased expression of HLA class II and CD86.
For any application
Application examples include flow cytometry or fluorescence-activated cell sorting, the study of T cell proliferation, apoptosis, and induction of anergy, study of antigen-specific T cells and regulation of T cell cytokine expression.
Polyclonal T-Cell Activation/Expansion
Isolated T cells are readily activated and/or expanded in culture with Dynabeads® Human T-Activator CD3/CD28 or Dynabeads® Mouse T-Activator CD3/CD28.
Antigen-Specific T-Cell Expansion
The new Dynabeads® T-Activator CD3/CD28/CD137 for expansion of human antigen-specific T cells and mouse antigen specific T cells are conjugated with anti-CD3, anti-CD28, and anti-CD137 antibodies.