TrueCut Cas9 is now available as a CTS-prototype for preclinical and translational research. Learn more »
Dynabeads™ Untouched™ Human CD8 T Cells Kit
Invitrogen™

Dynabeads™ Untouched™ Human CD8 T Cells Kit

The Dynabeads™ Untouched™ Human CD8 T Cells Kit is used to isolate pure and viable untouched CD8+ T cells fromRead more
Have Questions?
Catalog NumberQuantity
11348D1 kit
Catalog number 11348D
Price (USD)
1,160.00
Each
Quantity:
1 kit
Price (USD)
1,160.00
Each
The Dynabeads™ Untouched™ Human CD8 T Cells Kit is used to isolate pure and viable untouched CD8+ T cells from PBMC by negative isolation. The kit depletes CD4+ T cells, B cells, NK cells, monocytes, platelets, dendritic cells, granulocytes, and erythrocytes. The negatively isolated human CD8+ T-cells are left in the sample and have not been in contact with the Dynabeads™. The bead- and antibody-free CD8+ T cells are truly untouched, and in perfect shape for any functional assay or application.

Advantages of the Dynabeads™ Untouched™ Human CD8 T Cells Kit:

• High purity, recovery, and viability of human untouched CD8+ T cells
• Easy to use and to scale up
• No columns required

Magnetic bead-based Separation offers easy handling
An antibody mix towards the non-CD8 cells is added to the sample and allowed to bind to the cells. Dynabeads™ are added and will bind to the antibody-labeled cells during a short incubation. The bead-bound cells are quickly separated on a magnet and discarded. The remaining negatively isolated and untouched human CD8+ T cells can be directly analyzed in a flow cytometer and used in any application. For depletion or positive isolation of CD8+ cells, select from our range of Human CD8+ T Cell Isolation Products.

Learn more about Dynabeads™ products
• Find Dynabeads™ products for a whole range of applications.
• Find magnets for Dynabeads™ separations.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell TypeT Cells (CD8)
ClonalityMonoclonal
Final Product TypeCells
High-throughput CompatibilityNot High-throughput Compatible (Manual)
Isolation TechnologyNegative Isolation
No. of CellsProcesses ∼1 x 109 cells total
No. of Tests100 Isolations
Output Viability>95%, >98%
Product LineDYNAL, Dynabeads, Untouched
Purity or Quality GradeResearch Grade
Quantity1 kit
Sample TypePBMC
Shipping ConditionRoom Temperature
Starting Material Cell No.1 x 107 PBMCs per isolation
Target SpeciesHuman
Product TypeCell Isolation Kit
Unit SizeEach
Contents & Storage
The Kit Contains: 10 mL Depletion Dynabeads™ and 2 mL Antibody Mix with monoclonal antibodies towards human CD4, CD14, CD16a, CD16b, CD19, CD36, CD56, CD123 and CD235a (Glycophorin A).

Storage Conditions: 2°C to 8°C. Do not freeze.

Frequently asked questions (FAQs)

My Dynabeads magnetic beads are not pelleting well with the magnet. Do you have any suggestions for me?

Please review the following possibilities for why your Dynabeads magnetic beads are not pelleting:

- The solution is too viscous.
- The beads have formed aggregates because of protein-protein interaction.

Try these suggestions: - Increase separation time (leave tub on magnet for 2-5 minutes)
- Add DNase I to the lysate (~0.01 mg/mL)
- Increase the Tween 20 concentration to ~0.05% of the binding and/or washing buffer.
- Add up to 20 mM beta-merecaptoethanol to the binding and/or wash buffers.

Find additional tips, troubleshooting help, and resources within our Dynabeads Nucleic Acid Purification Support Center.

I have a long double-stranded DNA fragment I would like to isolate. What product do you recommend?

For biotin-labled DNA that is less than 1 kb, we recommend you use Dynabeads M270 Streptavidin and MyOne C1 magnetic beads. We recommend our Dynabeads KilobaseBINDER Kit, which is designed to immobilize long (>1 kb) double-stranded DNA molecules. The KilobaseBINDER reagent consists of M-280 Streptavidin-coupled Dynabeads magnetic beads along with a patented immobilization activator in the binding solution to bind to long, biotinylated DNA molecules for isolation. Please see the following link (https://www.thermofisher.com/us/en/home/life-science/dna-rna-purification-analysis/napamisc/capture-of-biotinylated-targets/immobilisation-of-long-biotinylated-dna-fragments.html) for more information in regards to long biotinylated DNA fragment isolation.

Find additional tips, troubleshooting help, and resources within our Dynabeads Nucleic Acid Purification Support Center.

Can I use Dynabeads magnetic beads to isolate single-stranded DNA templates?

Yes, Dynabeads magnetic beads can be used to isolate single-stranded DNA. Streptavidin Dynabeads magnetic beads can be used to target biotinylated DNA fragments, followed by denaturation of the double-stranded DNA and removal of the non-biotinylated strand. The streptavidin-coupled Dynabeads magnetic beads will not inhibit any enzymatic activity. This enables further handling and manipulation of the bead-bound DNA directly on the solid phase. Please see the following link (https://www.thermofisher.com/us/en/home/life-science/dna-rna-purification-analysis/napamisc/capture-of-biotinylated-targets/preparing-single-stranded-dna-templates.html) for more information in regards to single-stranded DNA capture.

Find additional tips, troubleshooting help, and resources within our Dynabeads Nucleic Acid Purification Support Center.

What is the magnetic susceptibility for Dynabeads magnetic beads?

Magnetic susceptibility is a measure of how quickly the beads will migrate to the magnet. This will depend on the iron content and the character of the iron oxide. The magnetic susceptibility given for the Dynabeads magnetic beads is the mass susceptibility, given either as cgs units/g or m^3/kg (the latter being an SI unit). For ferri- and ferromagnetic substances, the magnetic mass susceptibility is dependent upon the magnetic field strength (H), as the magnetization of such substances is not a linear function of H but approaches a saturation value with increasing field. For that reason, the magnetic mass susceptibility of the Dynabeads magnetic beads is determined by a standardized procedure under fixed conditions. The magnetic mass susceptibility given in our catalog is thus the SI unit. Conversion from Gaussian (cgs, emu) units into SI units for magnetic mass susceptibility is achieved by multiplying the Gaussian factor (emu/g or cgs/g) by 4 pi x 10^-3. The resulting unit is also called the rationalized magnetic mass susceptibility, which should be distinguished from the (SI) dimensionless magnetic susceptibility unit. In general, magnetic mass susceptibility is a measure of the force (Fz) influencing an object positioned in a nonhomogenous magnetic field. The magnetic mass susceptibility of the Dynabeads magnetic beads is measured by weighing a sample, and then subjecting the sample to a magnetic field of known strength. The weight (F1) is then measured, and compared to the weight of the sample when the magnetic field is turned off (F0). The susceptibility is then calculated as K x 10^-3 = [(F1-F0) x m x 0.335 x 10^6], where K is the mass susceptibility of the sample of mass m. The susceptibility is then converted to SI units.

Find additional tips, troubleshooting help, and resources within our Dynabeads Nucleic Acid Purification Support Center.

How can I determine coupling efficiency of Dynabeads magnetic beads?

There are different methods to check binding of ligands to the beads, including optical density (OD) measurement, fluorescent labeling, and radioactive labeling.

For OD measurement, you would measure the OD of the ligand before immobilization to the beads and compare it with the ligand concentration that is left in the supernatant after coating. This gives a crude measurement of how much protein has bound to the beads.

Protocol:

1.Set spectrophotometer to the right wavelength. As a blank, use the Coupling Buffer.
2.Measure the absorbance of the Pre-Coupling Solution. A further dilution may be necessary to read the absorbance, depending upon the amount of ligand added.
3.Measure the absorbance of the Post-Coupling Solution. A dilution may be necessary to read the absorbance.
4.Calculate the coupling efficiency, expressed as the % protein uptake, as follows. [(Pre-Coupling Solution x D) - (Post-Coupling Solution x D)] x 100/(Pre-Coupling Solution x D) where D = dilution factor.

For fluorescent labeling, we suggest negatively quantifying the amount of ligand bound by measuring ligand remaining in the coupling supernatant (compared to the original sample), rather than directly measuring the ligands on the beads. Add labeled ligand to the beads, and measure how much ligand is left in the supernatant (not bound to the beads). By comparing this with the total amount added in the first place, you can then calculate how much of the ligand that has been bound to the beads. Keep in mind that the Dynabeads magnetic beads are also autofluorescent, which is why direct measuring of fluorescence of the bead-bound ligands is not recommended, but rather this indirect approach. The label could be, for example, FITC/PE. Some researchers perform a direct approach with success (using a flow cytometer).

Radioactive labeling is the most sensitive method of the three, but it is also the most difficult one. It involves radioactively labeling a portion of the ligand. We use radiolabeled I-125 in tracer amounts and mix it with "cold" ligands in a known ratio before coupling. The absolute quantities for the ligand on the beads should be obtained by measuring the beads in a scintillation (gamma) counter and comparing the cpm with a standard.

Protocol:

1.Take out an appropriate amount of beads and wash the beads in 1 mL of binding buffer.
2.Pipette out desired amount of human IgG in a separate tube.
3.Mix the human IgG with I-125-labeled human IgG (30,000 - 100,000 cpm).
4.Dilute the mixture of human IgG and I-125-labeled human IgG to 100 mL in binding buffer.
5.Incubate for 30 minutes at room temperature and measure the cpm in a scintillation counter.
6.Wash the beads (with coating) four times, and measure cpm again.
The % binding is calculated by using the equation : (cpm after washing/cpm before washing)x100%.

Find additional tips, troubleshooting help, and resources within our Dynabeads Nucleic Acid Purification Support Center.