Overview
The Applied Biosystems QuantStudio 12K Flex Real-Time PCR System can simultaneously run up to four 3,072-reaction QuantStudio 12K Flex OpenArray plates in about four hours. When combined with the QuantStudio 12K Flex OpenArray AccuFill System, the QuantStudio 12K Flex system can produce up to 110,000 data points or more in an 8-hour day with as little as 20 minutes of hands-on time per run. A feature-rich touch screen interface and comprehensive software analysis tools for gene expression, and genotyping increases productivity.
Designed for the highest flexibility, the QuantStudio 12K Flex system has five-interchangeable block formats including: OpenArray plate, TaqMan Array card, 384-well, and 96-well (0.1 and 0.2 mL). This delivers unmatched versatility for the lab with flexible format requirements. The system also includes laboratory information management system (LIMS) support and features that assist with 21CFR Part 11 compliance are enabled for high-throughput, validated environments.
Features
The QuantStudio 12K Flex system offers:
Maximize throughput with minimal resources
Increase throughput by as much as 32-fold by running four OpenArray plates in a single run. 
Five interchangeable blocks
OpenArray plate, TaqMan Array Card, 384-well, 96-well (0.1 and 0.2 mL) blocks that are  interchangeable with no tools required
Automation compatibility
Increase productivity and reduce your manual tasks through automation compatibility with the Thermo Scientific Orbitor RS2 Microplate Mover
Maximum multiplexing & chemistry options
Proven fluorescence detection with enhanced OptiFlex system, allows up to six multiplexing targets 
Reduce start up time and hands on time
Start a 12,000-data-point experiment typically in 20 minutes or less. 
21CFR Part 11 compliance and LIMS support
Laboratory information management system (LIMS) support and features that assist with 21CFR Part 11 compliance are enabled for high-throughput, and an optional security, auditing, e-signature package can be added. 
Videos and demos
Scale experiments to meet the immediate throughput needs of your laboratory from 12K to a single reaction. Optional interchangeable blocks are available to run OpenArray plates, TaqMan Array cards, 384-well plates, 96-well plates (0.1 and 0.2 mL).
Start a 12,000 data point experiment in typically less than 20 minutes using automated sample loading with the QuantStudio 12K Flex AccuFill System.
Sample data
9-log separation
Real-time PCR reproducibility and 9-log dynamic range. Amplification plot shows results from real-time PCR of 18S DNA in 10-fold dilutions starting at 25,000 copies using the 384-well block. The data show highly reproducible results for input target quantities covering nine orders of magnitude (9 logs). 
Instrument calibration and verification kits
Maximize the performance of your QuantStudio 12K Flex system with the following calibration kits. 
QuantStudio 12K Flex system, 384-well, with QuantStudio Real-Time PCR Software v1.7 or earlier
Catalog # Name Size Price Qty
A24736 Each
208,00
4432271 Each
108,90
Online Exclusive
Ends 27-Mar-2026
198,00 Save 89,10 (45%)
4425559 Each
868,00
A24733 Each
208,00
4457334 Each
209,00
4432302 Each
193,00
4432308 Each
318,65
Online Exclusive
350,00 Save 31,35 (9%)
4432320 Each
216,70
Online Exclusive
Ends 27-Mar-2026
394,00 Save 177,30 (45%)
4432284 Each
109,45
Online Exclusive
Ends 27-Mar-2026
199,00 Save 89,55 (45%)
4432290 Each
230,00
4432296 Each
125,40
Special offer
Online exclusive
Ends 27-Mar-2026
228,00 Save 102,60 (45%)
4432278 Each
198,00
4432470 Each
3.380,00
4455280 Each
991,10
Special offer
Online exclusive
Ends 27-Mar-2026
1.802,00 Save 810,90 (45%)
QuantStudio 12K Flex system, 384-well, with QuantStudio Real-Time PCR Software v1.8 or later
Catalog # Name Size Price Qty
A26334 Each
310,20
Special offer
Online exclusive
Ends 27-Mar-2026
564,00 Save 253,80 (45%)
A26335 Each
308,00
Special offer
Online exclusive
Ends 27-Mar-2026
560,00 Save 252,00 (45%)
A26341 Each
618,20
Special offer
Online exclusive
Ends 27-Mar-2026
1.124,00 Save 505,80 (45%)
4432320 Each
216,70
Online Exclusive
Ends 27-Mar-2026
394,00 Save 177,30 (45%)
4455280 Each
991,10
Special offer
Online exclusive
Ends 27-Mar-2026
1.802,00 Save 810,90 (45%)
4425559 Each
868,00
QuantStudio 12K Flex system, 96-well, 0.1 mL, with QuantStudio Real-Time PCR Software v1.7 or earlier
QuantStudio 12K Flex system, 96-well, 0.1 mL, with QuantStudio Real-Time PCR Software v1.8 or later
Catalog # Name Size Price Qty
A26336 Each
194,15
Online Exclusive
Ends 27-Mar-2026
353,00 Save 158,85 (45%)
A26337 Each
194,15
Special offer
Online exclusive
Ends 27-Mar-2026
353,00 Save 158,85 (45%)
A26340 Each
351,00
A26342 Each
589,60
Online Exclusive
Ends 27-Mar-2026
1.072,00 Save 482,40 (45%)
4432426 Each
217,25
Online Exclusive
Ends 27-Mar-2026
395,00 Save 177,75 (45%)
4351979 Each
822,80
Online Exclusive
Ends 27-Mar-2026
1.496,00 Save 673,20 (45%)
4425618 Each
870,00
QuantStudio 12K Flex system, 96-well, 0.2 mL, with QuantStudio Real-Time PCR Software v1.7 or earlier
Catalog # Name Size Price Qty
A24738 Each
210,00
4432327 Each
107,80
Special offer
Online exclusive
Ends 27-Mar-2026
196,00 Save 88,20 (45%)
A24737 Each
209,00
4461599 Each
116,60
Special offer
Online exclusive
Ends 27-Mar-2026
212,00 Save 95,40 (45%)
4432358 Each
198,00
4432370 Each
189,75
Special offer
Online exclusive
Ends 27-Mar-2026
345,00 Save 155,25 (45%)
4432364 Each
217,80
Online Exclusive
Ends 27-Mar-2026
396,00 Save 178,20 (45%)
4432340 Each
198,00
4432346 Each
197,00
4432352 Each
107,80
Special offer
Online exclusive
Ends 27-Mar-2026
196,00 Save 88,20 (45%)
4432334 Each
107,80
Special offer
Online exclusive
Ends 27-Mar-2026
196,00 Save 88,20 (45%)
4432488 Each
3.385,00
4432382 Each
650,10
Online Exclusive
Ends 27-Mar-2026
1.182,00 Save 531,90 (45%)
QuantStudio 12K Flex system, 96-well, 0.2 mL, with QuantStudio Real-Time PCR Software v1.8 or later
Catalog # Name Size Price Qty
A26331 Each
199,10
Online Exclusive
Ends 27-Mar-2026
362,00 Save 162,90 (45%)
A26332 Each
194,15
Special offer
Online exclusive
Ends 27-Mar-2026
353,00 Save 158,85 (45%)
A26333 Each
194,70
Online Exclusive
Ends 27-Mar-2026
354,00 Save 159,30 (45%)
A26343 Each
614,90
Online Exclusive
Ends 27-Mar-2026
1.118,00 Save 503,10 (45%)
4432364 Each
217,80
Online Exclusive
Ends 27-Mar-2026
396,00 Save 178,20 (45%)
4432382 Each
650,10
Online Exclusive
Ends 27-Mar-2026
1.182,00 Save 531,90 (45%)
4425562 Each
381,70
Special offer
Online exclusive
Ends 27-Mar-2026
694,00 Save 312,30 (45%)
QuantStudio 12K Flex system, OpenArray
QuantStudio 12 Flex system, TaqMan Array Cards
Catalog # Name Size Price Qty
4432314 Each
5.570,00
4432250 Each
6.640,00
Specifications
* Optional software module available.
For Research Use Only. Not for use in diagnostic procedures.