MeltDoctor™ HRM Calibration Standard (20X)
Product Image
Applied Biosystems™

MeltDoctor™ HRM Calibration Standard (20X)

Applied Biosystems® MeltDoctor™ HRM Calibration Standard contains a DNA template and primers which, when mixed with PCR reagents and dye,Read more
Have Questions?
Catalog NumberQuantity
44255621 mL
Catalog number 4425562
Price (USD)
752.00
Each
Add to cart
Quantity:
1 mL
Price (USD)
752.00
Each
Add to cart
Applied Biosystems® MeltDoctor™ HRM Calibration Standard contains a DNA template and primers which, when mixed with PCR reagents and dye, can be used to prepare a plate for thermal and optical High Resolution Melt (HRM) calibration.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Calibrated DyeHRM (High Resolution Melting) Dye
Calibration TypeThermal Calibration, Optic Calibration
Concentration20 X
For Use With (Application)Real Time PCR (qPCR)
For Use With (Equipment)7500 Fast System, 7900HT Fast System
FormatTube
Product LineMeltDoctor
Quantity1 mL
Shipping ConditionWet Ice
Product TypeHRM Calibration Standard
Unit SizeEach
Contents & Storage
1 tube containing 1 mL of a 20X solution of DNA template and primers which, when mixed with PCR reagents and dye, can be used to prepare a calibration plate for thermal and optical High Resolution Melt (HRM).
Store at -15°C to -25°C

Guaranteed minimum shelf life is 60 days (exact expiry date printed on product and CofA).

Frequently asked questions (FAQs)

How long should the amplicon be for HRM analysis?

We recommend to use 50-250 bp long PCR amplicons. Typically, shorter amplicons can distinguish the genotypes for a SNP better, especially for Type III and Type IV SNPs. This is simply because a single base variation affects the melting behavior of a 100 bp amplicon stronger than of a 500 bp amplicon, for example. In longer fragments, the risk of covering multiple mutations is also increased.

What can be used as positive control reactions in HRM analysis?

Positive control reactions should contain template DNA with a known sequence. In SNP genotyping experiments, this could be a sample with a known genotype. Positive control(s) for all genotypes should be included where possible to serve as a reference in melting curve comparison and assigning genotypes for test samples. In mutation scanning experiments, a sample with a wild type sequence could serve as a positive control. The controls should preferably have the same DNA concentration as their corresponding test samples. Control DNA should also be eluted and/or diluted in the same buffer as the samples.

In HRM analysis, when is it better to use a 3-step cycling protocol rather than a 2-step protocol?

A 3-step cycling protocol is recommended for the analysis of complicated (especially Type IV SNP) targets, amplicons longer than 200 bp, and amplicons with a primer annealing temperature that is less than 60 degrees C.

The HRM software will not accept my calibration file. What is wrong?

Check that there are no outliers on the plate. You cannot omit any wells on the HRM calibration plate.

I have a 7900HT Fast Real-Time PCR System, and cannot open the data file in HRM software. What could be causing this?

There are a few possibilities. First, make sure the HRM Software version is v2.0.1, and the 7900HT Fast Real-Time PCR System software version is v2.3 or above. Second, check that the run method used was as recommended in the HRM protocol; make sure the ramp rate for the dissociation stage is 1%. Then try to open the calibration file from the HRM Software; if it does not open, the calibration file is defective. The defects could be due to a bad calibration plate or instrument uniformity issue.