Neural Stem Cells (NSCs)
Neural Stem Cells
Powering neural stem cell culture
Discover optimized solutions that can enable you to employ neural stem cells in your research.
What are neural stem cells?
Neural stem cells (NSCs) are a type of stem cell that can differentiate into the main types of cells found in the nervous system: neurons, astrocytes, and oligodendrocytes. These cells are capable of self-renewal and can generate the various cell types that make up the nervous system.
How are NSCs used in neuroscience research? 
NSCs are valuable tools in neuroscience research due to their ability to self-renew, differentiate into multiple neural cell types, and model various aspects of the nervous system. Their versatility and relevance to human biology make them an ideal tool for studying neural development, disease mechanisms, drug screening, and potential therapeutic applications. 
NSCs are being used to:
  • Understand how the nervous system develops and the role of different cell types.
  • Create models of neurological diseases such as Parkinson's, Alzheimer's, and multiple sclerosis to study disease mechanisms and test potential treatments.
  • Test the efficacy and safety of new drugs targeting neurological conditions.
  • Explore the potential to repair or replace damaged neural tissue in conditions like spinal cord injuries and stroke.
How are NSCs derived from iPSCs?
Neural stem cells (NSCs) can be generated from induced pluripotent stem cells (iPSCs) using a variety of methods that promote neural induction. The earliest developed methods relied on embryoid body (EB) formation and creation of neural rosettes. Although this process mimics early embryonic development, it is prone to heterogeneity and is a more time-consuming method. Methods leveraging monolayer culture and exposure to neural induction factors or dual-SMAD inhibition help provide a simpler and more homogenous approach. The neural induction factors that drive NSCs in monolayer culture can also be adapted to suspension cultures, allowing controlled neural induction in a 3D environment. The choice of method for generating NSCs from iPSCs depends on the specific needs of the research. 
Many NSC-based studies require efficient delivery of nucleic acids to modulate gene expression or introduce reporters. Chemical transfection reagents such as Lipofectamine reagents enable gentle delivery for sensitive NSC cultures, while electroporation-based approaches like the Neon NxT Electroporation System offer high efficiency for more demanding genetic manipulations.
Find the neural cell culture tools that are right for you
Having the right components for your complete neural cell culture medium is essential to achieving exceptional cell culture results. We offer researchers an array of neural stem cell products that support a multitude of established methods for derivation, expansion, differentiation, and analysis. You can choose based on your cell model needs.
Neural induction
Whether you need a simple kit or basal medium and supplement for complete medium that can be optimized, we have you covered with the key reagents to induce pluripotent stem cells into neural stem cells.
Catalog # Name Size Price Qty
A1647801 Each
20,930.00
線上優惠
Ends 30-Sep-2026
29,900.00 Save 8,970.00 (30%)
17504044 Each
4,655.00
線上優惠
Ends 30-Sep-2026
6,650.00 Save 1,995.00 (30%)
17504001 Each
48,640.00
線上優惠
Ends 30-Sep-2026
60,800.00 Save 12,160.00 (20%)
12587010 Each
5,460.00
線上優惠
Ends 30-Sep-2026
7,800.00 Save 2,340.00 (30%)
12587001 Each
***
-
11320033 Each
1,302.00
線上優惠
Ends 30-Sep-2026
1,860.00 Save 558.00 (30%)
11320082 Each
11,340.00
線上優惠
Ends 30-Sep-2026
16,200.00 Save 4,860.00 (30%)
21103049 Each
3,031.00
線上優惠
Ends 30-Sep-2026
4,330.00 Save 1,299.00 (30%)
17502048 Each
3,745.00
線上優惠
Ends 30-Sep-2026
5,350.00 Save 1,605.00 (30%)
17502001 Each
43,680.00
線上優惠
Ends 30-Sep-2026
54,600.00 Save 10,920.00 (20%)
NSC culture and proliferation
We offer several options for the growth and expansion of NSCs, most suitable option depends on how NSCs were derived.
Best for primary NSCs of human or rat
Learn more about StemPro NSC SFM—Serum-Free Human Neural Stem Cell Culture Medium
Product details: StemPro Neural Supplement
Best for NSCs derived from the PSC Neural Induction Medium
Learn more about neural induction and differentiation
Product details: PSC Neural Induction Medium, Neurobasal Medium
Best for dual SMAD methods
Catalog # Name Size Price Qty
17504044 Each
4,655.00
線上優惠
Ends 30-Sep-2026
6,650.00 Save 1,995.00 (30%)
17504001 Each
48,640.00
線上優惠
Ends 30-Sep-2026
60,800.00 Save 12,160.00 (20%)
12587010 Each
5,460.00
線上優惠
Ends 30-Sep-2026
7,800.00 Save 2,340.00 (30%)
12587001 Each
***
-
11320033 Each
1,302.00
線上優惠
Ends 30-Sep-2026
1,860.00 Save 558.00 (30%)
11320082 Each
11,340.00
線上優惠
Ends 30-Sep-2026
16,200.00 Save 4,860.00 (30%)
21103049 Each
3,031.00
線上優惠
Ends 30-Sep-2026
4,330.00 Save 1,299.00 (30%)
17502048 Each
3,745.00
線上優惠
Ends 30-Sep-2026
5,350.00 Save 1,605.00 (30%)
17502001 Each
43,680.00
線上優惠
Ends 30-Sep-2026
54,600.00 Save 10,920.00 (20%)
Differentiation from NSC to neurons
We offer several options to achieve efficient differentiation with varying levels of control and flexibility for optimization.
B-27 Supplement
A serum-free supplement designed to support the survival, growth, and maturation of neurons in vitro.
Learn more
Product details
B-27 Plus Neuronal Culture System
An optimized, serum-free culture system that enhances neuronal viability, synaptic activity, and long-term neuronal health.
Learn more
Product details
CultureOne Supplement
A defined supplement that selectively suppresses glial proliferation to promote enriched neuronal cultures.
Learn more
Product details
PSC Dopaminergic Neuron Differentiation kit
A complete, ready-to-use kit for efficiently differentiating human pluripotent stem cells into dopaminergic neurons.
Learn more
Product details
Neurobasal Medium
A basal medium formulated to support the growth and maintenance of primary and stem cell-derived neurons with reduced background activity.
Learn more
Product details
Genetic delivery for NSC workflows
In addition to optimized media and supplements, many NSC workflows require reliable delivery of DNA, RNA, or CRISPR components. Lipofectamine reagents support efficient, low-toxicity transfection in neural stem cells, while the Neon NxT Electroporation System provides scalable, high-performance delivery across a range of NSC formats and differentiation stages.
Cell analysis
Verifying neural stem cells (NSCs) and their progeny to confirm pluripotency or support differentiation into the desired cell type is a critical step in your research. The Human Neural Stem Cell Immunocytochemistry Kit provides a complete solution for image-based analysis of key NSC markers such as nestin, PAX6, SOX1, and SOX2, can help you confidently characterize NSCs and their derivatives.
Explore our portfolio of cell analysis, imaging, flow cytometry, and HCS/HCA solutions to streamline and scale your workflow.
Cell analysis instruments
Flow cytometry instruments
High-content screening (HCS) and high-content analysis (HCA)