Whole Allergen
d1 House dust mite
d1 House dust mite Scientific Information
Type:
Whole Allergen
Display Name:
House dust mite
Allergen code:
d1
Family:
Pyroglyphidae
Route of Exposure:
Inhalation
Latin Name:
Dermatophagoides pteronyssinus
Other Names:
House dust mite, Dust mite, European house dust mite
WHO/ICD-11 code:
XM8BV4
(ICD-11 is currently under implementation by WHO and the ICD-11 codes displayed in the encyclopedia may not yet be available in all countries)
Summary
DermatophagoidesSummary pteronyssinus (Der p) is a house dust mite specie originally found in house dust. This domestic mite is highly allergenic, and exposure to its allergens in sensitized individuals is one of the most common cause of respiratory allergic diseases in the world. The main exposure route is inhalation and symptoms typically affects the upper and lower airways and eyes, causing allergic rhinitis, asthma, and sometimes anaphylaxis. Exposure via the skin can cause atopic dermatitis. Several Der p allergens have been characterized and are present on airborne fecal and body particles. Extensive IgE cross-reactivity exists between Der p and other house dust mite species. Partial cross-reactivity has been demonstrated to storage mite species. Cross-reactivity also exists among allergens in Der p and related arthropods including crustaceans (shrimp, crab, and lobster) and insects (cockroach, grasshopper), as well as members of the mollusk phylum (snails, clams, oysters, and squid. Management of house dust mite allergy includes avoidance, pharmacotherapy and specific allergen immunotherapy (AIT).
Allergen
Nature
Unique characteristics of dust mites have allowed them to colonize the indoor environment in temperate regions of the world. Other traits facilitate the delivery of their allergens into close contact with skin and mucosal epithelium. House dust mites, including D pteronyssinus, produce a complex range of allergens and adjuvants which act together to induce both innate and adaptive immune reactions. Accordingly, the induction of allergy to these mites is an important cause of respiratory allergies, and the exposure of already sensitized individuals to mite products is a significant trigger of asthma exacerbations (Calderon 2015). 
Taxonomy
Dust mites belong to the phylum Arthropoda, class Arachnida. Within this phylum are 2 other classified groups which contain species having an important role in allergic disease, namely the class insects and subphylum crustacea (Miller 2019).
Some insects and arachnids are the source of important inhalant allergens, while some crustaceans are a major source of food allergens. During evolution, arthropods separated from other animals approximately 600 million years ago and the major classes were established within 100 million years.  Dust mites and insects such as cockroaches have therefore been separated for at least 400 million years (Miller 2019). 
Despite the huge number of arthropod species, only a few have been implicated as sources of indoor allergens and 90% of the related literature covers only five genera: Dermatophagoides, Blomia, Euroglyphus, Blatella and Periplaneta. Similarly, although multiple species of mites have been identified in indoor environments, only a few have been causally associated with allergic disease, namely D. pteronyssinus, D. farinae, Euroglyphus maynei and Blomia tropicalis (Miller 2019).
Tissue
Mite allergens come into contact with respiratory mucosa and skin via fecal particles, and to a much lesser extent via fragments of mite bodies. During its lifetime a mite produces approximately 1000 solid fecal waste particles of approximately 20 – 25 µ in diameter. This size of particle is capable of penetrating both upper and lower airways, thus coming into contact with lung mucosa as well as more superficial sites such as skin or eyes (Tovey 1981a, Tovey 1981b).
Mite fecal particles contain high concentrations of molecules, some which are allergenic proteins. More than 80% of mite allergen is on particles >10 μ and is undetectable in the air of undisturbed rooms. Fecal particles become transiently airborne during disturbance brought about by human activities such as sweeping, vacuuming or changing bedding. The particles are also an important source of other molecules, such as nucleic acids, that have adjuvant properties via interaction with toll-like receptors or pattern recognition receptors (PRRs) which recognize pathogen associated molecular patterns (PAMPS). Chitin, mite DNA, bacterial DNA, and endotoxin are known PAMPS found in house dust mite feces and bodies. The fecal particles can thus induce an IgE response but also contribute to non-allergen specific inflammation in the nose, lungs and skin  (Tovey 1981a, Tovey 1981b).
Epidemiology   
Worldwide distribution
Dust mites are found worldwide, except in cold, dry high-altitude regions, polar areas, and extremely arid desert zones. House dust mites thrive in temperate climates, particularly in households and dwellings with damp and humid conditions. Allergens from the species D. pteronyssinus and D. farinae are commonly found in dust samples from homes, indicating the presence of both species in the same environment. Within any given geographical area, dust mites vary in their distribution among specific regions and with the seasons (Zheng 2015). D. farinae can tolerate dryer environmental conditions. D. pteronyssinus tends to be more abundant in Europe than in the United States. In northern areas of Europe where the climate is extremely dry, dust mites may not survive the winter.
High altitude is generally inhospitable to house dust mites, which may partly explain why, before the discovery of dust mites, sanitaria for asthma and other respiratory diseases were often built at high altitudes, such as the Alps in Europe and Colorado in the USA. Dust mite growth (Vervloet 1982) and sensitization (Charpin 1991) are much lower in the Alps than at sea level, the apparent result of the lower indoor humidity at high altitude.
Living environment
Dust mites are most prevalent in households, where they thrive in dark environments and burrow deep into soft substrates like carpets, pillows, mattresses, and clothing (Schroer 2008). A higher mite allergen exposure has been correlated with higher education, higher household income, and lower population density in the home (Leaderer 2002). Mite allergen levels are reported higher in older homes and non-air conditioned dwellings (van Strien 2004). A study from China found significant enrichment of house dust mite major allergens in air-conditioner filters located in dining rooms, shopping malls, hotels, and households. The concentration of Der p 1 in the air increased notably one hour after the air-conditioners were turned on (Zhan 2015).
Route of Exposure         
Main route
House dust mite allergens trigger allergic reactions primarily through inhalation of mite fecal particles and body fragments, which become airborne after disturbances like vacuuming or making the bed. 
Other routes
Other routes include direct skin contact with contaminated bedding or furniture and accidental ingestion of mite particles by consumption of contaminated foods (Millner 2019).
Clinical Relevance
Allergic rhinitis
Significant proportions of the allergic rhinitis patients and chronic rhinosinusitis patients with allergy are sensitized to house dust mites (Rolla 2007). The link between allergen sensitization and symptoms can be demonstrated using nasal challenge with mite allergen, which produces obstruction and rhinorrhea that correlate with mite skin test reactivity (Chusakul 2010).
Information on the occurrence of ocular symptoms in association with allergic rhinitis is sparse, but one study of patients with allergic rhinitis to a variety of allergens including dust mites, found that most individuals also had ocular involvement with symptoms of pruritus, tearing, conjunctival injection, and eyelid edema (Klossek 2012).
Asthma
Dust mites including D pteronyssinus are one of the most frequent causes of respiratory allergies and mite exposure is a very important factor eliciting exacerbations of asthma (Calderon 2015). 
Mite allergen exposure as a trigger to exacerbate existing asthma has been clearly and repeatedly demonstrated (Miller 2019). The inhalation of dust mite allergen can have effects beyond bronchospasm, decreasing mucociliary clearance and thus increasing the deposition of other inhaled particles. The combination of mite sensitization and mite exposure together correlate with the severity of asthma symptoms (Tunnicliffe 1999), with increased exhaled nitric oxide and bronchial hyper-reactivity (Langley 2003) and with acute exacerbations resulting in admission to hospital (Sporik 1993).
Sensitization to Dermatophagoides mites also appears to exacerbate asthma attacks in children suffering from rhinovirus infections (Soto-Quiros 2012). The addition of viral infection to allergen exposure in mite-sensitized individuals results in more severe attacks with acute wheezing (Soto-Quiros 2012) and hospitalization (Green 2002, Murray 2006).
Mite allergy is a major risk factor for asthma and mite sensitization early in life has a significant impact on subsequent pulmonary function. One multicenter, birth cohort study followed 1314 children from birth to 13 years of age  (Illi 2006). Asthma symptoms and lung function, specific IgE, and perennial allergen exposure (mite, cat and dog dander) were assessed at regular intervals. The great majority (90%) of children with wheeze but no sensitization had lost their symptoms by school age and retained normal lung function at puberty. In contrast, sensitization to perennial allergens including mite, which developed in the first 3 years of life was correlated with compromised lung function at school age.  Sensitization and exposure occurring later than 3 years of age resulted in much weaker effects on lung function, and sensitization to seasonal allergens such as pollens had no effect on subsequent lung function (Illi 2006).
Atopic Dermatitis
The prevalence of sensitization to mites can be very high in patients with atopic dermatitis. The increase in the permeability of atopic skin and the ability of mite proteases to decrease skin barrier function may allow more effective sensitization with aeroallergens, initiating a cycle of inflammation and further allergen exposure (Miller 2019).
Other diseases
Systemic allergic symptoms, sometimes severe, can occur after inadvertent ingestion of dust mites present in a food that has been colonized by mites, referred to as oral mite anaphylaxis. This was first reported in 1993, in a patient who ate a fried pastry made from flour that had been contaminated with D. farinae mites (Erben 1993). Subsequently, multiple cases of systemic allergy have been reported in mite sensitized patients who ate a variety of foods made with mite-contaminated foods including pancakes (Wen 2005), wheat and corn flour (Blanco 1997, Guerra Bernd 2001), grits (Posthumus 2012) and pizza (Kymioni 2025). Storing any opened packages of baked goods mixes, grains, or flour in a refrigerator will prevent the growth of mite populations.
Diagnostic Relevance
In vitro diagnostics
Mite allergy diagnosis involves a physical examination and medical history review to identify associations between mite exposure and symptoms. Confirmation of Der p sensitization is achieved through in vitro specific IgE testing and skin prick testing. Specific IgE testing may be preferred over skin prick testing for individuals taking medications that could interfere with skin test results, such as certain antihistamines, or for those with skin conditions like dermatitis.
A systematic review and meta-analyses of eight studies involving 1,095 patients supported the use of Der p 1 and Der p 2 IgE as useful tools to aid in the diagnosis of Der p allergic rhinitis and/or allergic asthma, detecting sensitization in approximately 80-90% of patients who are allergic to house dust mites (Tian 2017).
Prevention and Therapy
Prevention strategies
Avoidance
The effects of house dust mite reduction and avoidance measures on asthma, rhinitis, and eczema, have been assessed in systematic reviews following the Cochran Review methodology and criteria. In the Cochrane avoidance review on asthma, several trials were included involving in total 1,421 participants (Zuiani 2020). The review on rhinitis included 9 randomized controlled trials involving 501 participants (Sheikh 2010) and for eczema, 7 randomized controlled studies were included in the final review (Nankervis 2015).
Prevention of sensitization
Current Cochrane review evidence does not strongly support the effectiveness of avoidance measures in preventing sensitization to house dust mites. Zuiani (2020) reports that some studies have shown significant reductions in asthma exacerbations and symptoms with multi-faceted allergen avoidance approaches. However, these findings are more related to symptom reduction rather than the prevention of sensitization. Similarly, Sheikh (2010) concluded that while acaricides and extensive bedroom-based environmental control programs might reduce symptoms of allergic rhinitis, there is no strong evidence to support their effectiveness in preventing sensitization. Nankervis (2015) also reported that there is no clear evidence that house dust mite reduction and avoidance measures can prevent sensitization to house dust mites. However, the Nankervis 2015 review indicates that trials examining this question have been small and of low quality, making it difficult to draw definitive conclusions.
Reduction of allergic symptoms
In Zuiani (2020) several studies are reported to have shown significant reductions in asthma exacerbations, symptoms and acute health care visits after multi-faceted avoidance approaches targeting multiple allergens, including house dust mites. Effective strategies for allergen avoidance, particularly for house dust mites, include using mite-impermeable bedding covers, vacuum cleaners with HEPA filters, extensive bedroom-based environmental control programs, education on allergen avoidance and smoke exposure reduction, and combining physical barriers, acaricides, laundry services, and tailored education. Overall, a multi-faceted set of measures tailored to the patient's characteristics and home exposures was recommended to achieve the greatest reduction in allergen exposure and improving asthma symptoms.
The evidence supporting the effectiveness of house dust mite avoidance measures in managing perennial allergic rhinitis is limited (Sheikh 2010). Some studies indicate that acaricides and comprehensive bedroom-based environmental control programs may alleviate rhinitis symptoms. However, the Cochrane review highlights the need for more high-quality research to confirm these findings and provide definitive recommendations for the use of house dust mite reduction and avoidance measures in managing perennial allergic rhinitis.
The Cochrane evidence regarding the effectiveness of house dust mite reduction and avoidance measures in managing eczema was found limited and inconclusive (Nankervis 2015). Seven randomized controlled trials involving 324 adults and children with eczema were reviewed, including interventions like mattress and bedding covers, high-quality vacuuming, and mite-killing sprays. The trials did not provide sufficient evidence to recommend any specific measures, with no significant clinical benefit observed. The Cochrane review highlights the need for high-quality, long-term trials of simple interventions to assess the effectiveness of house dust allergen reduction and avoidance in managing eczema.
Allergen immunotherapy
Specific allergen immunotherapy (AIT) is a treatment option for house dust allergic patients with allergic rhinitis and allergic asthma. AIT is available in two main forms: subcutaneous immunotherapy (SCIT) administered via injections and sublingual immunotherapy (SLIT) taken under the tongue (Muraro 2018, Creticos 2024).
A recent real-world evidence study involving 7,260 patients with house dust mite AIT prescriptions and 21,780 control patients demonstrated that subcutaneous AIT reduces the need for medication to treat allergic rhinitis and allergic asthma. Additionally, it was found to delay the onset of asthma medication in patients with allergic rhinitis (Mösges 2024).
In the past decade, sublingual AIT tablets targeting house dust mite allergies have been developed and received regulatory approvals (Creticos 2021). These AIT tablets have demonstrated efficacy and safety in treating house dust mite induced allergic rhinitis in adults (Klimek 2016, Demoly 2021) and children (Sasamoto 2024), as well as adult asthma (Canonica 2016).
A systematic review including 23 randomized controlled trials with 1, 957 patients across 13 countries evaluated the effectiveness of sublingual and subcutaneous AIT for atopic dermatitis (AD). The findings suggest that AIT likely improves AD severity, itch, and sleep disturbance, with moderate-to-high certainty evidence for several outcomes. SCIT was associated with a modest increase in adverse events, while SLIT had fewer side effects. The review highlights the potential benefits of AIT for long-term AD control and calls for further research to optimize treatment protocols and understand long-term impacts (Yepes-Nuñez 2023).
Component-resolved diagnosis (CRD) using major allergens Der p 1 and Der p 2 may allow the identification of those patients who are best suited for house dust mite AIT immunotherapy (Pittner 2004) and a diagnostic algorithm integrating house dust mite allergenic molecules has been developed (Dramburg 2023).
Specific IgG4 to induced by house dust mite AIT can be useful to confirm successful immunological stimulation against Der p molecules (Zhao 2016) and delivery of AIT earlier in life may be associated with a greater increase in Der p specific IgG4 (Lai 2013). However, there is currently no firm clinical evidence that levels of specific IgG4 can predict or serve as biomarker for clinical efficacy in individual patients receiving AIT (Agache 2019).
Molecular Aspects         
The proteomes of house dust mites have been characterized identifying Der p specific proteins and potential allergens (Waldron 2019).
Allergenic molecules                                                                                                                                                          
To date, a total of 35 allergens for D. pteronyssinus have been identified and formally named by the WHO/IUIS Allergen Nomenclature Sub-Committee, see table 1.
Der p 1 and Der p 2 are major allergens in D pteronyssinus. The sensitization pattern in a total of 1,302 Dermatophagoides-allergic patients from Europe (466 adults and 423 children), Canada (350 adults), USA (35 adults) and Japan (28 adults) showed that >70% and >80% of patients were sensitized to house dust mite group 1 and group 2 allergens, respectively, from D. pteronyssinus and/or D. Farinae species (Batard 2016).
Furthermore, 20–47% of patients also had IgE to mite allergens from groups 4, 5, 7, 13, 15, 21, and 23. All patients had IgE to allergens present in mite bodies and faecal particles (Batard 2016).
Der p 1 is a 25 kDa glycoprotein allergen with cysteine protease activity, belonging to the papain family. The molecular structures of recombinant Der p 1 with its propeptide (Meno 2005) and a fully mature form of Der p 1 have been reported (de Halleux 2006). Der p 1 and Der f 1 share 81% amino acid sequence identity and similar three-dimensional structures (Chruszcz 2009) leading to IgE cross-reactivity.
Der p 2 is a heat and pH stable protein of 14 kDa, belonging to the NPC2 cholesterol-binding protein family. The molecular structures of recombinant Der p 2 (Derewenda 2002) and Der f 2 (Johannessen 2005) reveal that these allergens share highly similar three-dimensional structures leading to the high degree of IgE cross-reactivity between Der p 2 and Der f 2 (Yasueda 1989).
Sensitization to mites often starts at an early age and early sensitization to Der p 1, Der p 2 and Der p 23 have been found associated with asthma development (Posa 2017). Asthmatic atopic patients are also reported to be sensitized to a wider range of mite allergens than atopic patients without asthma and have higher levels of IgE specific for each allergen (Resch 2015).
Allergen
Biological function
Molecular weight (kDa)
Der p 1
Cysteine protease
24
Der p 2
NPC2 family; MD-2-related lipid recognition (ML) domain containing protein
15
Der p 3
Trypsin
31
Der p 4
Alpha-amylase
60
Der p 5
Group 5/21 allergen
14
Der p 6
Chymotrypsin
25
Der p 7
Bactericidal permeability-increasing like protein
26,30 and 31
Der p 8
Glutathione S-transferase
27
Der p 9
Collagenolytic serine protease
29
Der p 10
Tropomyosin
36
Der p 11
Paramyosin
103
Der p 13
Cytosolic Fatty Acid Binding Protein
15
Der p 14
Apolipophorin
177
Der p 15
Chitinase, contains a C-terminal peritrophin-A-like domain
58.8 and 61.4
Der p 16
Gelsolin
55
Der p 18
Non-catalytic chitinase-like protein (contains a C-terminal peritrophin-A-like domain)
49.2
Der p 20
Arginine kinase
40
Der p 21
Group 5/21 allergen
14.726
Der p 23
Peritrophin-like protein domain (PF01607)
14 (dimer)
Der p 24
Ubiquinol-cytochrome c reductase binding protein; Cytochrome b-c1 complex subunit 7
13
Der p 25
Triosphosphate isomerase
27
Der p 26
Myosin light-chain
14.1
Der p 27
Serpin serine protease inhibitor
45
Der p 28
Heat shock protein Hsp70
45
Der p 29
Cyclophilin
27.7
Der p 30
Ferritin
12.1
Der p 31
Cofilin
16.8
Der p 32
Inorganic pyrophosphatase
45
Der p 33
alpha-tubulin
44.2
Der p 36
C2 domain containing protein
23
Der p 37
Chitin binding protein (contains 2 peritrophin-A-like domains)
30
Der p 38
Bacteriolytic enzyme
15
Der p 39
Troponin C
18
Der p 40
Thioredoxin like protein
12
Table 1. Current List of Dermatophagoides pteronyssinus allergens from WHO/IUIS Allergen Nomenclature Sub-Committee.
Molecular spreading
A study which followed a large cohort of children and tested for IgE to D. pteronyssinus extract and 12 individual allergens at regular intervals found that at age 20 years, the most frequent IgE specificity was for Der p 2, Der p 1, and Der p 23 (group A molecules; prevalence, >40%), followed by Der p 5, Der p 7, Der p 4, and Der p 21 (group B molecules; prevalence, 15% to 30%) and Der p 11, Der p 18, clone 16, Der p 14, and Der p 15 (group C molecules; prevalence, <10%).  The IgE response specificity started almost invariably with group A molecules and broadened sequentially first to group B and finally to group C molecules. A broader polymolecular IgE sensitization pattern was associated with early onset of IgE sensitization, parental hay fever, and higher exposure to mites. Individuals with the broadest IgE sensitization stage (i.e. ABC) had a significantly higher risk of mite-related allergic rhinitis and asthma than unsensitized participants. The presence of IgE to Der p 1 or Der p 23 at age 5 years or less predicted asthma at school age (Posa 2017).
Cross-reactivity
Studies have demonstrated almost complete IgE cross-reactivity between extracts of Der p and Der f and partial or low cross-reactivity between Der p and storage mites Lepidoglyphus destructor, Glycyphagus domesticus, Tyrophagus putrescentiae, Acarus siro and Blomia tropicalis (Pittner 2004, Zhang 2012). The group 1 and group 2 major allergens of Der p of Der f show high degree of IgE cross-reactivity.
Cross-reactivity also exists among allergens in dust mites and related arthropods including crustaceans (shrimp, crab, and lobster) and insects (cockroach, grasshopper), as well as members of the mollusk phylum (snails, clams, oysters, and squid). A pan-allergen responsible for these IgE cross-reactions to invertebrates is the muscle protein tropomyosin (Reese 1999, Sidenius 2001). In D. pteronyssinus mites, tropomyosin is represented by Der p 10 which shares sequence homology with shrimp tropomyosin Pen a 1, American cockroach tropomyosin Per a 7, and lobster tropomyosin Hom a 1 (Ayuso 2002). However, it is likely that tropomyosin is not the only relevant cross-reacting allergen, as there exist dust mite sensitive patients with shrimp allergy but not snail allergy, or with snail allergy but not shrimp allergy ​(Bessot 2010).
Explained Results
Allergen information
House dust mite Der p is an important indoor source of allergens, thriving in humid environments, beds and carpets. Allergens are present on mite fecal and body particles. Der p 1 and Der p 2 are the major allergens of Der p. Close to forty Der p allergens have been characterized.
Clinical relevance
Specific IgE to Der p is associated with allergic rhinitis, asthma, atopic dermatitis, and less frequently with symptoms following the intake of food contaminated by mites.
Cross reactivity
Extensive IgE cross-reactivity exists between extracts of Der p and Der f and partial or low cross-reactivity between Der p and storage mites. Cross-reactivity also exists among allergens in dust mites and related arthropods including crustaceans (shrimp, crab, and lobster) and insects (cockroach, grasshopper), as well as members of the mollusk phylum (snails, clams, oysters, and squid.
Compiled by: Dr. Christian Fischer, October 2020.
Last reviewed by Dr. Michael Spangfort, September 2025
References
Agache I, Lau S, Akdis CA, Smolinska S, Bonini M, Cavkaytar O, Flood B, Gajdanowicz P, Izuhara K, Kalayci O, Mosges R, Palomares O, Papadopoulos NG, Sokolowska M, Angier E, Fernandez-Rivas M, Pajno G, Pfaar O, Roberts GC, Ryan D, Sturm GJ, van Ree R, Varga EM, van Wijk RG, Yepes-Nuñez JJ, Jutel M. EAACI Guidelines on Allergen Immunotherapy: House dust mite-driven allergic asthma. Allergy. 2019;74(5):855-873.
Ayuso R, Reese G, Leong-Kee S, Plante M, Lehrer SB. Molecular basis of arthropod cross-reactivity: IgE-binding cross-reactive epitopes of shrimp, house dust mite and cockroach tropomyosins. Int Arch Allergy Immunol. 2002;129(1):38-48.
Batard T, Baron-Bodo V, Martelet A, Le Mignon M, Lemoine P, Jain K, et al. Patterns of IgE sensitization in house dust mite-allergic patients: implications for allergen immunotherapy. Allergy. 2016;71(2):220-9.
Bessot JC, Metz-Favre C, Rame JM, De Blay F, Pauli G. Tropomyosin or not tropomyosin, what is the relevant allergen in house dust mite and snail cross allergies? Eur Ann Allergy Clin Immunol. 2010;42(1):3-10.
Blanco C, Quiralte J, Castillo R, Delgado J, Arteaga C, Barber D, et al. Anaphylaxis after ingestion of wheat flour contaminated with mites. J Allergy Clin Immunol. 1997;99(3):308-13.
Calderon MA, Kleine-Tebbe J, Linneberg A, De Blay F, Hernandez Fernandez de Rojas D, Virchow JC, et al. House Dust Mite Respiratory Allergy: An Overview of Current Therapeutic Strategies. J Allergy Clin Immunol Pract. 2015;3(6):843-55.
Canonica GW, Virchow JC, Zieglmayer P, Ljørring C, Smith IM, Mosbech H. Efficacy and safety of SQ house dust mite (HDM) SLIT-tablet treatment of HDM allergic asthma. Expert Rev Clin Immunol. 2016;12(8):805-15.
Charpin D, Birnbaum J, Haddi E, Genard G, Lanteaume A, Toumi M, et al. Altitude and allergy to house-dust mites. A paradigm of the influence of environmental exposure on allergic sensitization. Am Rev Respir Dis. 1991;143(5 Pt 1):983-6.
Chruszcz M, Chapman MD, Vailes LD, Stura EA, Saint-Remy JM, Minor W, Pomés A. Crystal structures of mite allergens Der f 1 and Der p 1 reveal differences in surface-exposed residues that may influence antibody binding. J Mol Biol. 2009;386(2):520-30.
Chusakul S, Phannaso C, Sangsarsri S, Aeumjaturapat S, Snidvongs K. House-dust mite nasal provocation: a diagnostic tool in perennial rhinitis. Am J Rhinol Allergy. 2010;24(2):133-6.
Creticos PS. New insights in mite immunotherapy - sublingual tablets. Curr Opin Allergy Clin Immunol. 2021;21(6):602-610.
Creticos PS, Gunaydin FE, Nolte H, Damask C, Durham SR. Allergen Immunotherapy: The Evidence Supporting the Efficacy and Safety of Subcutaneous Immunotherapy and Sublingual Forms of Immunotherapy for Allergic Rhinitis/Conjunctivitis and Asthma. J Allergy Clin Immunol Pract. 2024, 12(6):1415-1427.
de Halleux S, Stura E, VanderElst L, Carlier V, Jacquemin M, Saint-Remy JM. Three-dimensional structure and IgE-binding properties of mature fully active Der p 1, a clinically relevant major allergen. J Allergy Clin Immunol. 2006; 117(3):571-6.
Demoly P, Corren J, Creticos P, De Blay F, Gevaert P, Hellings P, Kowal K, Le Gall M, Nenasheva N, Passalacqua G, Pfaar O, Tortajada-Girbés M, Vidal C, Worm M, Casale TB. A 300 IR sublingual tablet is an effective, safe treatment for house dust mite-induced allergic rhinitis: An international, double-blind, placebo-controlled, randomized phase III clinical trial. J Allergy Clin Immunol. 2021;147(3):1020-1030.
Derewenda U, Li J, Derewenda Z, Dauter Z, Mueller GA, Rule GS, Benjamin DC. The crystal structure of a major dust mite allergen Der p 2, and its biological implications. J Mol Biol. 2002;318(1):189-97.
Dramburg S, Hilger C, Santos AF, de Las Vecillas L, Aalberse RC, Acevedo N, Aglas L, Altmann F, Arruda KL, Asero R, Ballmer-Weber B, Barber D, Beyer K, Biedermann T, Bilo MB, Blank S, Bosshard PP, Breiteneder H, Brough HA, Bublin M, Campbell D, Caraballo L, Caubet JC, Celi G, Chapman MD, Chruszcz M, Custovic A, Czolk R, Davies J, Douladiris N, Eberlein B, Ebisawa M, Ehlers A, Eigenmann P, Gadermaier G, Giovannini M, Gomez F, Grohman R, Guillet C, Hafner C, Hamilton RG, Hauser M, Hawranek T, Hoffmann HJ, Holzhauser T, Iizuka T, Jacquet A, Jakob T, Janssen-Weets B, Jappe U, Jutel M, Kalic T, Kamath S, Kespohl S, Kleine-Tebbe J, Knol E, Knulst A, Konradsen JR, Korošec P, Kuehn A, Lack G, Le TM, Lopata A, Luengo O, Mäkelä M, Marra AM, Mills C, Morisset M, Muraro A, Nowak-Wegrzyn A, Nugraha R, Ollert M, Palosuo K, Pastorello EA, Patil SU, Platts-Mills T, Pomés A, Poncet P, Potapova E, Poulsen LK, Radauer C, Radulovic S, Raulf M, Rougé P, Sastre J, Sato S, Scala E, Schmid JM, Schmid-Grendelmeier P, Schrama D, Sénéchal H, Traidl-Hoffmann C, Valverde-Monge M, van Hage M, van Ree R, Verhoeckx K, Vieths S, Wickman M, Zakzuk J, Matricardi PM, Hoffmann-Sommergruber K. EAACI Molecular Allergology User's Guide 2.0. Pediatr Allergy Immunol. 2023;34 Suppl 28:e13854.
Erben AM, Rodriguez JL, McCullough J, Ownby DR. Anaphylaxis after ingestion of beignets contaminated with Dermatophagoides farinae. J Allergy Clin Immunol. 1993;92(6):846-9.
Green RM, Custovic A, Sanderson G, Hunter J, Johnston SL, Woodcock A. Synergism between allergens and viruses and risk of hospital admission with asthma: case-control study. BMJ. 2002;324(7340):763.
Guerra Bernd LA, Arruda LK, Barros Antunes HB. Oral anaphylaxis to mites. Allergy. 2001;56(1):83-4.
Illi S, von Mutius E, Lau S, Niggemann B, Gruber C, Wahn U, et al. Perennial allergen sensitisation early in life and chronic asthma in children: a birth cohort study. Lancet. 2006;368(9537):763-70.
Johannessen BR, Skov LK, Kastrup JS, Kristensen O, Bolwig C, Larsen JN, Spangfort M, Lund K, Gajhede M. Structure of the house dust mite allergen Der f 2: implications for function and molecular basis of IgE cross-reactivity. FEBS Lett. 2005;579(5):1208-12.
Klimek L, Mosbech H, Zieglmayer P, Rehm D, Stage BS, Demoly P. SQ house dust mite (HDM) SLIT-tablet provides clinical improvement in HDM-induced allergic rhinitis. Expert Rev Clin Immunol. 2016;12(4):369-77.
Klossek JM, Annesi-Maesano I, Pribil C, Didier A. The burden associated with ocular symptoms in allergic rhinitis. Int Arch Allergy Immunol. 2012;158(4):411-7.
Kymioni VM, Karatsoli I, Koronas N, Karagiorga I, Kakleas K. Oral mite anaphylaxis after pizza consumption in a child: A case report and review of the literature. Allergol Immunopathol (Madr). 2025;53:149-152.
Lai X, Li J, Xiao X, Liu E, Zhang C, Wang H, Gjesing B, Zhong N, Spangfort MD. Specific IgG4 production during house dust mite immunotherapy among age, gender and allergic disease populations. Int Arch Allergy Immunol. 2013;160(1):37-46.
Langley SJ, Goldthorpe S, Craven M, Morris J, Woodcock A, Custovic A. Exposure and sensitization to indoor allergens: association with lung function, bronchial reactivity, and exhaled nitric oxide measures in asthma. J Allergy Clin Immunol. 2003;112(2):362-8.
Leaderer BP, Belanger K, Triche E, Holford T, Gold DR, Kim Y, et al. Dust mite, cockroach, cat, and dog allergen concentrations in homes of asthmatic children in the northeastern United States: impact of socioeconomic factors and population density. Environ Health Perspect. 2002;110(4):419-25.
Meno K, Thorsted PB, Ipsen H, Kristensen O, Larsen JN, Spangfort MD, Gajhede M, Lund K. The crystal structure of recombinant proDer p 1, a major house dust mite proteolytic allergen. J Immunol. 2005;175(6):3835-45.
Miller JD. The Role of Dust Mites in Allergy. Clin Rev Allergy Immunol. 2019;57(3):312-29.
Muraro A, Roberts G, Halken S, Agache I, Angier E, Fernandez-Rivas M, Gerth van Wijk R, Jutel M, Lau S, Pajno G, Pfaar O, Ryan D, Sturm GJ, van Ree R, Varga EM, Bachert C, Calderon M, Canonica GW, Durham SR, Malling HJ, Wahn U, Sheikh A. EAACI guidelines on allergen immunotherapy: Executive statement. Allergy. 2018;73(4):739-743.
Murray CS, Poletti G, Kebadze T, Morris J, Woodcock A, Johnston SL, et al. Study of modifiable risk factors for asthma exacerbations: virus infection and allergen exposure increase the risk of asthma hospital admissions in children. Thorax. 2006;61(5):376-82.
Mösges R, Richter H, Sager A, Weber J, Müller T. House dust mite immunotherapy: A real-world, prescription data-based analysis. Clin Transl Allergy. 2024;14(7):e12382.
Nankervis H, Pynn EV, Boyle RJ, Rushton L, Williams HC, Hewson DM, Platts-Mills T. House dust mite reduction and avoidance measures for treating eczema. Cochrane Database Syst Rev. 2015;1(1):CD008426.
Pittner G, Vrtala S, Thomas WR, Weghofer M, Kundi M, Horak F, et al. Component-resolved diagnosis of house-dust mite allergy with purified natural and recombinant mite allergens. Clin Exp Allergy. 2004;34(4):597-603.
Posa D, Perna S, Resch Y, Lupinek C, Panetta V, Hofmaier S, et al. Evolution and predictive value of IgE responses toward a comprehensive panel of house dust mite allergens during the first 2 decades of life. Journal of Allergy and Clinical Immunology. 2017;139(2):541-549.
Posthumus J, Borish L. A 71-year-old man with anaphylaxis after eating grits. Allergy Asthma Proc. 2012;33(1):110-3.
Reese G, Ayuso R, Lehrer SB. Tropomyosin: an invertebrate pan-allergen. Int Arch Allergy Immunol. 1999;119(4):247-58.
Resch Y, Michel S, Kabesch M, Lupinek C, Valenta R, Vrtala S. Different IgE recognition of mite allergen components in asthmatic and nonasthmatic children. J Allergy Clin Immunol. 2015;136(4):1083-1091.
Rolla G, Guida G, Heffler E, Badiu I, Bommarito L, De Stefani A, et al. Diagnostic Classification of Persistent Rhinitis and Its Relationship to Exhaled Nitric Oxide and Asthma: A Clinical Study of a Consecutive Series of Patients. Chest. 2007;131(5):1345-1352.
Sasamoto K, Nagakura KI, Asaumi T, Fusayasu N, Ohashi-Doi K, Yanagida N, Sato S, Ebisawa M. Efficacy and safety of sublingual immunotherapy using house dust mite tablet for 1-4 years old children with perennial allergic rhinitis. Pediatr Allergy Immunol. 2024;35(7):e14203.
Sheikh A, Hurwitz B, Nurmatov U, van Schayck CP. House dust mite avoidance measures for perennial allergic rhinitis. Cochrane Database Syst Rev. 2010;2010(7):CD001563.
Sidenius KE, Hallas TE, Poulsen LK, Mosbech H. Allergen cross-reactivity between house-dust mites and
other invertebrates. Allergy. 2001;56(8):723-733.
Soto-Quiros M, Avila L, Platts-Mills TA, Hunt JF, Erdman DD, Carper H, et al. High titers of IgE antibody to dust mite allergen and risk for wheezing among asthmatic children infected with rhinovirus. J Allergy Clin Immunol. 2012;129(6):1499-1505.
Sporik R, Platts-Mills TA, Cogswell JJ. Exposure to house dust mite allergen of children admitted to hospital with asthma. Clinical & Experimental Allergy. 1993;23(9):740-746.
Tian M, Zhou Y, Zhang W, Cui Y. Der p 1 and Der p 2 specific immunoglobulin E measurement for diagnosis of Dermatophagoides pteronyssinus allergy: A systematic review and meta-analysis. Allergy Asthma Proc. 2017;38(5):333-342.
Tovey ER, Chapman MD, Platts-Mills TAE. Mite faeces are a major source of house dust allergens. Nature. 1981a;289(5798):592-593.
Tovey ER, Chapman MD, Wells CW, Platts-Mills TA. The distribution of dust mite allergen in the houses of patients with asthma. Am Rev Respir Dis. 1981b;124(5):630-635.
Tunnicliffe WS, Fletcher TJ, Hammond K, Roberts K, Custovic A, Simpson A, et al. Sensitivity and exposure to indoor allergens in adults with differing asthma severity. Eur Respir J. 1999;13(3):654-659.
Vervloet D, Penaud A, Razzouk H, Senft M, Arnaud A, Boutin C, et al. Altitude and house dust mites. Journal of Allergy and Clinical Immunology. 1982;69(3):290-296.
Waldron R, McGowan J, Gordon N, McCarthy C, Mitchell EB, Doyle S, Fitzpatrick DA. Draft Genome Sequence of Dermatophagoides pteronyssinus, the European House Dust Mite. Genome Announc. 2017;5(32):e00789-17.
Wen DC, Shyur SD, Ho CM, Chiang YC, Huang LH, Lin MT, et al. Systemic anaphylaxis after the ingestion of pancake contaminated with the storage mite Blomia freemani. Ann Allergy Asthma Immunol. 2005;95(6):612-4.
Yasueda H, Mita H, Yui Y, Shida T. Comparative analysis of physicochemical and immunochemical properties of the two major allergens from Dermatophagoides pteronyssinus and the corresponding allergens from Dermatophagoides farinae. Int Arch Allergy Appl Immunol. 1989;88(4):402-407.
Yepes-Nuñez JJ, Guyatt GH, Gómez-Escobar LG, Pérez-Herrera LC, Chu AWL, Ceccaci R, Acosta-Madiedo AS, Wen A, Moreno-López S, MacDonald M, Barrios M, Chu X, Islam N, Gao Y, Wong MM, Couban R, Garcia E, Chapman E, Oykhman P, Chen L, Winders T, Asiniwasis RN, Boguniewicz M, De Benedetto A, Ellison K, Frazier WT, Greenhawt M, Huynh J, Kim E, LeBovidge J, Lind ML, Lio P, Martin SA, O'Brien M, Ong PY, Silverberg JI, Spergel J, Wang J, Wheeler KE, Schneider L, Chu DK. Allergen immunotherapy for atopic dermatitis: Systematic review and meta-analysis of benefits and harms. J Allergy Clin Immunol. 2023;151(1):147-158.
Zhan X, Li C, Xu H, Xu P, Zhu H, Diao J, Li N, Zhao B. Air-conditioner filters enriching dust mites allergen. Int J Clin Exp Med. 2015;8(3):4539-4544.
Zhang C, Li J, Lai X, Zheng Y, Gjesing B, Spangfort MD, Zhong N. House dust mite and storage mite IgE reactivity in allergic patients from Guangzhou, China. Asian Pac J Allergy Immunol. 2012;30(4):294-300.
Zhao D, Lai X, Tian M, Jiang Y, Zheng Y, Gjesing B, Zhong N, Spangfort MD. The Functional IgE-Blocking Factor Induced by Allergen-Specific Immunotherapy Correlates with IgG4 Antibodies and a Decrease of Symptoms in House Dust Mite-Allergic Children. Int Arch Allergy Immunol. 2016;169(2):113-120.
Zheng YW, Lai XX, Zhao DY, Zhang CQ, Chen JJ, Zhang L, Wei QY, Chen S, Liu EM, Norback D, Gjesing B, Zhong NS, Spangfort DM. Indoor Allergen Levels and Household Distributions in Nine Cities Across China. Biomed Environ Sci. 2015;28(10):709-717.
Zuiani C, Custovic A. Update on House Dust Mite Allergen Avoidance Measures for Asthma. Curr Allergy Asthma Rep. 2020;20(9):50.
van Strien RT, Gehring U, Belanger K, Triche E, Gent J, Bracken MB, et al. The influence of air conditioning, humidity, temperature and other household characteristics on mite allergen concentrations in the northeastern United States. Allergy. 2004;59(6):645-52.
van der Heide S, De Monchy JG, De Vries K, Dubois AE, Kauffman HF. Seasonal differences in airway hyperresponsiveness in asthmatic patients: relationship with allergen exposure and sensitization to house dust mites. Clin Exp Allergy. 1997;27(6):627-33.