Whole Allergen
e201 Canary bird feathers
e201 Canary bird feathers Scientific Information
Type:
Whole Allergen
Display Name:
Canary bird feathers
Allergen code:
e201
Family:
Fringillidae
Latin Name:
Serinus canarius
Other Names:
Canary
WHO/ICD-11 code:
XE69N
(ICD-11 is currently under implementation by WHO and the ICD-11 codes displayed in the encyclopedia may not yet be available in all countries)
Environmental Characteristics
Other topics
The Canary is one of the most common domestic birds, together with Budgerigars and Parrots. In 1994, these birds were estimated at 25 million in US households, and more than 8 millions in German ones (1).
 These birds descend from the wild Serin Finch or the very similar Wild Canary, and their ancestors were brought from the Canary Islands, Madeira, or the Azores. The Wild Canary, which still exists, is brownish green and looks like a Sparrow. Captive breeding has gone on for 500 years, during most of which time Canaries were the most popular cage birds in the world, because of their singing and mimicry and the ease with which they breed in captivity. The yellow mutation (with the bird remaining the size and shape of a Finch) has been the most popular, but Canaries are available in many colors and a range of sizes, shapes, patterns and songs. The German (or Hartz Mountain) Roller is the classic song Canary and was the most numerous Canary breed in the world for many years. The American Singer is a newer song breed that was developed in the United States. Usually somewhat aloof in nature, they do not typically bond to people
Clinical Relevance
IgE-mediated reactions
Asthma, allergic rhinitis and allergic conjunctivitis may result following exposure to Canary feathers, epithelial cells or droppings (2). The allergic manifestations may present as Bird Fancier´s Asthma and as Bird-Egg Syndrome, with symptoms such as rhinitis, urticaria and angioedema (3)(2) and also as gastro-intestinal problems (4). Specific IgE has been found in patients exposed to Canary (1, 3, 4).
In a study, IgE antibodies to Canary were present in about 25% of the bird fanciers investigated (2). Among 212 sera from Budgerigar and Canary fanciers with symptoms of rhinitis and/or bronchial asthma, in 25 of 98 Canary feather-specific IgE antibody measurements, a significant level of specific IgE was found. In 3 sera IgE antibodies against sera from both birds were present. Canary feathers were shown to contain IgE-binding antigens that were not present in the bird sera and droppings (5).
In a prospective study, 258 adults were investigated for sensitisation against bird antigens (Budgerigar, Canary, Pigeon) using an intracutaneous test. Thirty (38%) showed a positive skin reaction with at least 1 of the 3 extracts tested. In persons not exposed to birds, 24/96 (25%) were found to react to at least one of the three allergen extracts (6).
Extrinsic allergic alveolitis, also known as hypersensitivity pneumonitis, Bird Fancier's Lung and Farmer's Lung, is a disease of inflammation of the lung parenchyma in the terminal bronchioles and alveoli. Symptoms may start soon after exposure to bird allergens or after many years, and may include breathlessness, cough, occasional chills, and fever. Death may also result.
The disease occurs after exposure to organic dust, especially after close contact with Pigeons or other birds such as Budgerigars, Parrots, Canaries, Parakeets, Cockatiels, Doves and Finches. Exposure results in the development of immunoglobulin antibodies including IgE (1), IgM (8), IgA and various IgG subclasses (7-9). The antibodies may be found in the sera and saliva of patients (10) as well as in the sera of asymptomatic but exposed subjects (11).
The allergenic proteins may be found in bird serum, droppings, skin scales, feathers and, in the case of Pigeons, Pigeon bloom (a waxy fine dust which coats the feathers of Pigeons). Contact may result from handling birds, cleaning their lofts, or exposure to the organic dust drifting down from a ceiling or roof where birds nest.This is typified in a report of a 54-year-old man who presented with features consistent with extrinsic allergic alveolitis occurring after contact with his pet birds (12).Diagnosis is based on a characteristic clinical picture and a typical x-ray pattern, accompanied by the presence of specific IgG antibodies (13).The measurement of specific IgG using IgG tracer technology has been shown to be a sensitive and specific assay for the routine diagnostic testing of extrinsic allergic alveolitis (14).
Molecular Aspects
Well-defined major allergenic bands with molecular mass of 20-30 kDa and 67 kDa have been detected and identified in IgE immunoblots with feather extracts as well as with serum proteins of Budgerigar, Parrot, Pigeon, Canary, and Hen. Inhalable feather dust was shown to contain several allergenic components which cross-react with serum allergens/antigens of the same as well as of other bird species (1)(15). The allergens have not been fully characterised yet.
Cross-reactivity
As noted above, inhalable feather dust contains several allergenic components which cross-react with serum allergens/antigens of the same as well as of other bird species (1).
Cross-reactivity between Canary and other phylogenetically related bird species may be expected, and in Canary-allergic patients, significant IgE antibody titers to Parrot, Budgerigar, Chicken, Pigeon, Goose and Duck have been reported (3, 4),  en in patients without known exposure1. Moreover, cross-reactivity to Hen's egg, Bird-Egg Syndrome, should be considered,. Here the livetins present in egg yolk have been suggested as the major cross-reacting allergen (4-5). This adult type of egg intolerance must be distinguished from the common egg-white allergy of atopic children
Compiled By
Last reviewed- April 2022
References
  1. Tauer-Reich I, Fruhmann G, Czuppon AB, Baur X. Allergens causing bird fancier's asthma. Allergy. 1994;49(6):448-53.
  2. van Toorenenbergen AW, Gerth van Wijk R, van Dooremalen G, Dieges PH. Immunoglobulin E antibodies against budgerigar and canary feathers. Int Arch Allergy Appl Immunol. 1985;77(4):433-7.
  3. de Maat-Bleeker F, van Dijk AG, Berrens L. Allergy to egg yolk possibly induced by sensitization to bird serum antigens. Annals of allergy. 1985;54(3):245-8.
  4. van Toorenenbergen AW, Huijskes-Heins MI, Gerth van Wijk R. Different pattern of IgE binding to chicken egg yolk between patients with inhalant allergy to birds and food-allergic children. Int Arch Allergy Immunol. 1994;104(2):199-203.
  5. van Toorenenbergen AW, Gerth van Wijk R, van Dooremalen G, Dieges PH. Immunoglobulin E antibodies against budgerigar and canary feathers. Int Arch Allergy Appl Immunol 1985;77(4):433-7
  6. Bosenecker A, Musken H, Bergmann KC. Sensitization in budgerigar owners. [German] Pneumologie 1998;52(4):209-13
  7. Baldwin CI, Todd A, Bourke SJ, Allen A, Calvert JE. IgG subclass responses to pigeon intestinal mucin are related to development of pigeon fanciers' lung. Clinical and experimental allergy : journal of the British Society for Allergy and Clinical Immunology. 1998;28(3):349-57.
  8. Todd A, Coan R, Allen A. Pigeon breeders' lung; IgG subclasses to pigeon intestinal mucin and IgA antigens. Clinical and experimental immunology. 1993;92(3):494-9.
  9. Yoshizawa Y, Miyashita Y, Inoue T, Sumi Y, Miyazaki Y, Sato T, et al. Sequential evaluation of clinical and immunological findings in hypersensitivity pneumonitis: serial subclass distribution of antibodies. Clinical immunology and immunopathology. 1994;73(3):330-7.
  10. McSharry C, MacLeod K, McGregor S, Speekenbrink AB, Sriram S, Boyd F, et al. Mucosal immunity in extrinsic allergic alveolitis: salivary immunoglobulins and antibody against inhaled avian antigens among pigeon breeders. Clinical and experimental allergy : journal of the British Society for Allergy and Clinical Immunology. 1999;29(7):957-64.
  11. Rodrigo MJ, Benavent MI, Cruz MJ, Rosell M, Murio C, Pascual C, et al. Detection of specific antibodies to pigeon serum and bloom antigens by enzyme linked immunosorbent assay in pigeon breeder’s disease. Occup Environ Med 200057(3):159-64
  12. Sutton PP, Pearson A, du Bois RM. Canary fancier's lung. Clin Allergy 1984;14(5):429-31
  13. Rodriguez de Castro F, Carrillo T, Castillo R, Blanco C, Diaz F, Cuevas M. Relationship between characteristics of exposure to pigeon antigens. Clinical manifestations and humoral immune response. Chest 1993;103(4):1059-63
  14. Lopata A, Schinkel M, Andersson C, Johansson G, van Hage-Hamsten M. Quantification of IgG antibodies to bird antigens in the diagnosis of extrinsic allergic alveolitis (EAA) using the UniCAP system. (Manuscript in preparation)
  15. Bosenecker A, Musken H, Bergmann KC. Sensitization in budgerigar owners. [German] Pneumologie 1998;52(4):209-13
  16.